Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen Sequence: MPLPVALQTR LAKRGILKHL EPEPEEEIIA EDYDDDPVDY EATRLEGLPP SWYKVFDPSC GLPYYWNADT DLVSWLSPHD PNSVVTKSAK KLRSSNADAE EKLDRSHDKS DRGHDKSDRS HEKLDRGHDK SDRGHDKSDR DRERGYDKVD RERERDRERD RDRGYDKADR EEGKERRHHR REELAPYPKS KKAVSRKDEE LDPMDPSSYS DAPRGTWSTG LPKRNEAKTG ADTTAAGPLF QQRPYPSPGA VLRANAEASR TKQQD
PQBP1 is a nuclear polyglutamine-binding protein that contains a WW domain. Mutations in this gene are associated with X-linked mental retardation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 38 kDa nuclear protein containing a WW domain; Npw38; Polyglutamine tract-binding protein 1; Polyglutamine-binding protein 1; PQBP-1; scurfy candidate 2
Gene Aliases: Npw38; PQBP-1; Pqbp1; Sfc2
UniProt ID: (Rat) Q6PCT5, (Mouse) Q91VJ5
Entrez Gene ID: (Rat) 302557, (Mouse) 54633
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support