Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PRDM2 Antibodies

          Proteintech

          PRDM2 Polyclonal Antibody

          View all (10) PRDM2 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite PRDM2 Polyclonal Antibody

          • Antibody Testing Data (1)
          PRDM2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PRDM2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PRDM2 Antibody (27718-1-AP) in WB

          HeLa cells were subjected to SDS PAGE followed by western blot with 27718-1-AP (PRDM2 antibody) at dilution of 1:300 incubated at room temperature for 1.5 hours. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PRDM2 Antibody in Western Blot (WB)

          Product Details

          27718-1-AP

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:200-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PRDM2 Fusion Protein Ag26769 (142-264 aa encoded by NM_001007257)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.47 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Wet ice (domestic); Dry ice (international)

          Product Specific Information

          Immunogen sequence: MGEDNPEIA AAIEEERASA RSKRSSPKSR KGKKKSQENK NKGNKIQDIQ LKTSEPDFTS ANMRDSAEGP KEDEEKPSAS ALEQPATLQE VASQEVPPEL ATPAPAWEPQ PEPDERLEAA ACEVN (142-264 aa encoded by NM_001007257)

          Target Information

          PRDM2, a member of nuclear/histone methyltransferase superfamily, consists of an N-terminal PR domain, an Rb-binding motif, 8 Kruppel-like zinc finger motifs, Nuclear hormone-receptor binding motif and a C-terminal PR-domain binding motif. Ubiquitously expressed in low levels, PRDM2 exists in 2 isoforms, a 280 kDa RIZ1 containing PR-domain and a 250 kDa truncated RIZ2 that lacks PR-domain. A tumor suppressor with a prominent role in cell growth and differentiation, PRDM2 binds to Retinoblastoma protein, estrogen receptor, GATA-3 and TPA-responsive element (MTE) of Heme-oxygenase-1 gene and influences pathogenesis of Retinoblastoma, transcriptional regulation during neuronal differentiation, transactivation of Heme-oxygenase-1 gene and specific effector of estrogen action. It is inactivated or altered due to mutations and hypermethylation in many types of human cancers suggesting a potential biomarker.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: G3b; GATA-3 binding protein G3B; GATA-3-binding protein G3B; Lysine N-methyltransferase 8; MTB-ZF; MTBzf; MTE-binding protein; PR domain containing 2, with ZNF domain; PR domain zinc finger protein 2; PR domain-containing protein 2; retinoblastoma protein-binding zinc finger protein; Retinoblastoma protein-interacting zinc finger protein; RP5-1177E19.1; Zinc finger protein RIZ; zinc-finger DNA-binding protein

          View more View less

          Gene Aliases: HUMHOXY1; KMT8; MTB-ZF; PRDM2; RIZ; RIZ1; RIZ2

          View more View less

          UniProt ID: (Human) Q13029

          View more View less

          Entrez Gene ID: (Human) 7799

          View more View less

          Function(s)
          DNA binding transcription factor activity, sequence-specific DNA binding zinc ion binding histone-lysine N-methyltransferase activity
          Process(es)
          transcription, DNA-templated regulation of transcription, DNA-templated determination of adult lifespan response to estradiol histone lysine methylation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0