Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • AMPK gamma-2 Antibodies

          Abnova

          PRKAG2 Polyclonal Antibody, MaxPab™

          View all (8) AMPK gamma-2 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite PRKAG2 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          PRKAG2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PRKAG2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PRKAG2 Antibody (H00051422-B01P) in WB

          Western Blot analysis of PRKAG2 expression in transfected 293T cell line by PRKAG2 MaxPab polyclonal antibody. Lane 1: PRKAG2 transfected lysate(36.08 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PRKAG2 Antibody in Western Blot (WB)

          Product Details

          H00051422-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PRKAG2 (NP_077747.1, 1 a.a. approximately 328 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MLEKLEFEDE AVEDSESGVY MRFMRSHKCY DIVPTSSKLV VFDTTLQVKK AFFALVANGV RAAPLWESKK QSFVGMLTIT DFINILHRYY KSPMVQIYEL EEHKIETWRE LYLQETFKPL VNISPDASLF DAVYSLIKNK IHRLPVIDPI SGNALYILTH KRILKFLQLF MSDMPKPAFM KQNLDELGIG TYHNIAFIHP DTPIIKALNI FVERRISALP VVDESGKVVD IYSKFDVINL AAEKTYNNLD ITVTQALQHR SQYFEGVVKC NKLEILETIV DRIVRAEVHR LVVVNEADSI VGIISLSDIL QALILTPAGA KQKETETE

          Target Information

          AMP-activated protein kinase (AMPK) is a heterotrimeric protein composed of a catalytic alpha subunit, a noncatalytic beta subunit, and a noncatalytic regulatory gamma subunit. Various forms of each of these subunits exist, encoded by different genes. AMPK is an important energy-sensing enzyme that monitors cellular energy status and functions by inactivating key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. This gene is a member of the AMPK gamma subunit family and encodes a protein with four cystathionine beta-synthase domains. Mutations in this gene have been associated with ventricular pre-excitation (Wolff-Parkinson-White syndrome), progressive conduction system disease and cardiac hypertrophy. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 5'-AMP-activated protein kinase subunit gamma-2; ampk gamma; AMPK gamma2; AMPK subunit gamma-2; H91620p; OTTHUMP00000211956; protein kinase, AMP-activated, gamma 2 non-catalytic subunit

          View more View less

          Gene Aliases: AAKG; AAKG2; CMH6; H91620p; PRKAG2; WPWS

          View more View less

          UniProt ID: (Human) Q9UGJ0

          View more View less

          Entrez Gene ID: (Human) 51422

          View more View less

          Function(s)
          AMP-activated protein kinase activity cAMP-dependent protein kinase inhibitor activity ATP binding cAMP-dependent protein kinase regulator activity phosphorylase kinase regulator activity AMP binding protein kinase binding protein kinase activator activity ADP binding
          Process(es)
          glycogen metabolic process regulation of glycolytic process protein phosphorylation negative regulation of protein kinase activity fatty acid biosynthetic process ATP biosynthetic process carnitine shuttle cell cycle arrest positive regulation of peptidyl-threonine phosphorylation sterol biosynthetic process macroautophagy regulation of fatty acid metabolic process activation of protein kinase activity intracellular signal transduction regulation of fatty acid biosynthetic process positive regulation of protein kinase activity regulation of fatty acid oxidation regulation of glucose import negative regulation of protein serine/threonine kinase activity regulation of signal transduction by p53 class mediator
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-9267x:80/100.66.134.224:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0