Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSSGTMKFNG YLRVRIGEAV GLQPTRWSLR HSLFKKGHQL LDPYLTVSVD QVRVGQTSTK QKTNKPTYNE EFCANVTDGG HLELAVFHET PLGYDHFVAN CTLQFQELLR TTGASDTFEG WVDLEPEGKV FVVITLTGSF TEATLQRDRI FKHFTRKRQR AMRRRVHQIN GHKFMATYLR QPTYCSHCRE FIWGVFGKQG YQCQVCTCVV HKRCHHLIVT ACTCQNNINK VDSKIAEQRF GINIPHKFSI HNYKVPTFCD HCGSLLWGIM RQGLQCKICK MNVHIRCQAN VAPNCGVNAV ELAKTLAGMG LQPGNISPTS KLVSRSTLRR QGKESSKEGN GIGVNSSNRL GIDNFEFIRV LGKGSFGKVM LARVKETGDL YAVKVLKKDV ILQDDDVECT MTEKRILSLA RNHPFLTQLF CCFQTPDRLF FVMEFVNGGD LMFHIQKSRR FDEARARFYA AEIISALMFL HDKGIIYRDL KLDNVLLDHE GHCKLADFGM CKEGICNGVT TATFCGTPDY IAPEILQEML YGPAVDWWAM GVLLYEMLCG HAPFEAENED DLFEAILNDE VVYPTWLHED ATGILKSFMT KNPTMRLGSL TQGGEHAILR HPFFKEIDWA QLNHRQIEPP FRPRIKSRED VSNFDPDFIK EEPVLTPIDE GHLPMINQDE FRNFSYVSPE LQP
Protein kinase C (PKC) is a family of serine- and threonine-specific protein kinases that can be activated by calcium and second messenger diacylglycerol. PKC family members phosphorylate a wide variety of protein targets and are known to be involved in diverse cellular signaling pathways. PKC family members also serve as major receptors for phorbol esters, a class of tumor promoters. Each member of the PKC family has a specific expression profile and is believed to play a distinct role in cells.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: MGC26269; MGC5363; nPKC-eta; PKC-L; Protein kinase C eta type; protein kinase C, eta
Gene Aliases: nPKC-eta; PKC-L; PKCL; PRKCH; PRKCL
UniProt ID: (Human) P24723
Entrez Gene ID: (Human) 5583
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support