Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Sequence of this protein is as follows: MAEVTREGFR LKKILIEDWK KAREEKQKKL IELQAGKKSL EDQVEMLRTV KEEAEKPERE AKEQHQKLWE EQLAAAKAQQ EQELAADAFK ELDDDMDGTV SVTELQTHPE LDTDGDGALS EAEAQALLSG DTQTDATSFY DRVWAAIRDK YRSEALPTDL PTPSAPDLTE PKEEQPPVPS SPTEEEEEEE EEEEEAEEEE EEEDSEEAPP PLSPPQPASP AEEDKMPPYD EQTQAFIDAA QEARNKFEEA ERSLKDMEES IRNLEQEISF DFGPNGEFAY LYSQCYELTT NEYVYRLCPF KLVSQKPKLG GSPTSLGTWG SWIGPDHDKF SAMKYEQGTG CWQGPNRSTT VRLLCGKETM VTSTTEPSRC EYLMELMTPA ACPEPPPEAP TEDDHDEL
SET domain-containing proteins, such as SMYD2, catalyze lysine methylation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 80K-H protein; advanced glycation end-product receptor 2; AGE-binding receptor 2; GLU2B; Glucosidase 2 subunit beta; glucosidase II; Glucosidase II subunit beta; hepatocystin; PKCSH; Protein kinase C substrate 60.1 kDa protein heavy chain; protein kinase C substrate, 80 Kda protein
Gene Aliases: AGE-R2; G19P1; GIIB; PCLD; PKCSH; PLD1; PRKCSH; VASAP-60
UniProt ID: (Human) P14314
Entrez Gene ID: (Human) 5589
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support