Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MPSRTGPKME GSGGRVRLKA HYGGDIFITS VDAATTFEEL CEEVRDMCRL HQQHPLTLKW VDSEGDPCTV SSQMELEEAF RLARQCRDEG LIIHVFPSTP EQPGLPCPGE DKSIYRRGAR RWRKLYRANG HLFQAKRFNR RAYCGQCSER IWGLARQGYR CINCKLLVHK RCHGLVPLTC RKHMDSVMPS QEPPVDDKNE DADLPSEETD GIAYISSSRK HDSIKDDSED LKPVIDGMDG IKISQGLGLQ DFDLIRVIGR GSYAKVLLVR LKKNDQIYAM KVVKKELVHD DEDIDWVQTE KHVFEQASSN PFLVGLHSCF QTTSRLFLVI EYVNGGDLMF HMQRQRKLPE EHARFYAAEI CIALNFLHER GIIYRDLKLD NVLLDADGHI KLTDYGMCKE GLGPGDTTST FCGTPNYIAP EILRGEEYGF SVDWWALGVL MFEMMAGRSP FDIITDNPDM NTEDYLFQVI LEKPIRIPRF LSVKASHVLK GFLNKDPKER LGCRPQTGFS DIKSHAFFRS IDWDLLEKKQ ALPPFQPQIT DDYGLDNFDT QFTSEPVQLT PDDEDAIKRI DQSEFEGFEY INPLLLSTEE SV
The PKC family of serine/threonine kinases, including PRKCZ (PKC zeta), is activated intracellularly by signal transduction pathways. In humans, at least 12 different PKC polypeptides have been identified. These isoforms differ in primary structure, tissue distribution, subcellular localization, mode of action in vitro, response to extracellular signals, and substrate specificity. PKC alpha, beta I, beta II and gamma form the conventional family; their activities are Ca2+- and phospholipid-dependent. PKC zeta plays a key regulatory role in a variety of cellular functions including cell growth and differentiation, gene expression, hormone secretion and membrane function.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: kinase nPKC-zeta; KPCZ; nPKC-zeta; nPKC-zeta PRKCZ; Protein kinase C zeta type; protein kinase C, zeta
Gene Aliases: PKC-ZETA; PKC2; PRKCZ
UniProt ID: (Human) Q05513
Entrez Gene ID: (Human) 5590
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support