Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MEAERRRQAE KPKKGRVGSS LLPERHPATG TPTTTVDSSA PPCRRLPGAG GGRSRFSPQG GQRGRPHSRR RHRTTFSPVQ LEQLESAFGR NQYPDIWARE SLARDTGLSE ARIQVWFQNR RAKQRKQERS LLQPLAHLSP AAFSSFLPES TACPYSYAAP PPPVTCFPHP YSHALPSQPS TGGAFALSHQ SEDWYPTLHP APAGHLPCPP PPPMLPLSLE PSKSWN
PROP1 encodes a paired-like homeodomain transcription factor in the developing pituitary gland. Expression occurs prior to and is required for expression of pou domain transcription factor 1, which is responsible for pituitary development and hormone expression. Mutations in this gene have been associated with combined pituitary hormone deficiency-2 as well as deficiencies in luteinizing hormone, follicle-stimulating hormone, growth hormone, prolactin, and thyroid-stimulating hormone.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Homeobox protein prophet of Pit-1; Pituitary-specific homeodomain factor; PROP-1; prophet of Pit1, paired-like homeodomain transcription factor
Gene Aliases: CPHD2; PROP-1; PROP1
UniProt ID: (Human) O75360
Entrez Gene ID: (Human) 5626
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support