Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LKLAMDLKGK MPNKMIRKGV FKDQHFDQNL NFMYIEVDKV TEREKVTVMS TINPTKDLLA DLIGCQRLPE DQRKKVHQLK DLLDQILMLD PAKRISINQA LQHAFIQE
Pre-mRNA splicing occurs in two sequential transesterification steps, and the protein encoded by this gene is thought to be involved in pre-mRNA splicing and in signal transduction. This protein belongs to a kinase family that includes serine/arginine-rich protein-specific kinases and cyclin-dependent kinases. This protein is regarded as a CDK-like kinase with homology to mitogen-activated protein kinases.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: dJ1013A10.1 (PRP4 protein kinase homolog); protein-serine/threonine kinase; PRP4 kinase; PRP4 pre-mRNA processing factor 4 homolog B; PRP4 pre-mRNA-processing factor 4 homolog; RP5-1013A10.2; Serine/threonine-protein kinase PRP4 homolog
Gene Aliases: dJ1013A10.1; KIAA0536; PR4H; PRP4; PRP4B; PRP4H; PRP4K; PRPF4K
UniProt ID: (Human) Q13523
Entrez Gene ID: (Human) 8899
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support