Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Cell lysate).
Immunogen sequence: YAEPTVLKWI RENIPEWKNC IIVSPDAGGA KRVTSIADQL NVDFALIHKE RKKANEVDCI VLVGDVNDRV AILVDDMADT CVTICLAADK LLSAGATR
Catalyzes the synthesis of phosphoribosylpyrophosphate (PRPP) that is essential for nucleotide synthesis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hCG_1643027; Phosphoribosyl pyrophosphate synthase 1-like 1; Phosphoribosyl pyrophosphate synthase III; phosphoribosylpyrophosphate synthetase subunit III; PRPS1-like 1; PRS-III; ribose-phosphate diphosphokinase catalytic chain III; Ribose-phosphate pyrophosphokinase 3; ribose-phosphate pyrophosphokinase III
Gene Aliases: 1700011K15Rik; PRPS1; PRPS1L1; PRPS3; PRPSL; PRS-III
UniProt ID: (Human) P21108
Entrez Gene ID: (Human) 221823, (Rat) 298954, (Mouse) 75456
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support