Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Sequence of this protein is as follows: MAHRPPSPAL ASVLLALLLS GAARAAEIVG GHEAQPHSRP YMASLQMRGN PGSHFCGGTL IHPSFVLTAA HCLRDIPQRL VNVVLGAHNV RTQEPTQQHF SVAQVFLNNY DAENKLNDIL LIQLSSPANL SASVATVQLP QQDQPVPHGT QCLAMGWGRV GAHDPPAQVL QELNVTVVTF FCRPHNICTF VPRRKAGICF GDSGGPLICD GIIQGIDSFV IWGCATRLFP DFFTRVALYV DWIRSTLRRV EAKGRP
Serine protease that degrades elastin, fibronectin, laminin, vitronectin, and collagen types I, III, and IV (in vitro) (PubMed:3198760, PubMed:2033050, PubMed:28240246). By cleaving and activating receptor F2RL1/PAR-2, enhances endothelial cell barrier function and thus vascular integrity during neutrophil transendothelial migration (PubMed:23202369). May play a role in neutrophil transendothelial migration, probably when associated with CD177 (PubMed:22266279). [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: AGP7; azurophil granule protein 7; C-ANCA antigen; Leukocyte proteinase 3; Myeloblastin; Neutrophil proteinase 4; NP-4; P29; PR-3; Proteinase 3; serine proteinase, neutrophil; Wegener autoantigen; Wegener granulomatosis autoantigen
Gene Aliases: ACPA; AGP7; C-ANCA; CANCA; MBN; MBT; mPR3; NP-4; NP4; P29; PR-3; PR3; PRTN3
UniProt ID: (Human) P24158, (Mouse) Q61096
Entrez Gene ID: (Human) 5657, (Mouse) 19152
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support