Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PSD Antibodies

          Abnova

          PSD Polyclonal Antibody, MaxPab™

          View all (5) PSD antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite PSD Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          PSD Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PSD Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PSD Antibody (H00005662-B01P) in WB

          Western Blot analysis of PSD expression in transfected 293T cell line by PSD MaxPab polyclonal antibody. Lane1:PSD transfected lysate(112.64 KDa). Lane2:Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PSD Antibody in Western Blot (WB)

          Product Details

          H00005662-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -

          Immunocytochemistry (ICC/IF)

          10-100 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Rat

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PSD (NP_002770.3, 1 a.a. approximately 1024 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5-1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAQGAMRFCS EGDCAISPPR CPRRWLPEGP VPQSPPASMY GSTGSLLRRV AGPGPRGREL GRVTAPCTPL RGPPSPRVAP SPWAPSSPTG QPPPGAQSSV VIFRFVEKAS VRPLNGLPAP GGLSRSWDLG GVSPPRPTPA LGPGSNRKLR LEASTSDPLP ARGGSALPGS RNLVHGPPAP PQVGADGLYS SLPNGLGGPP ERLATLFGGP ADTGFLNQGD TWSSPREVSS HAQRIARAKW EFFYGSLDPP SSGAKPPEQA PPSPPGVGSR QGSGVAVGRA AKYSETDLDT VPLRCYRETD IDEVLAEREE ADSAIESQPS SEGPPGTAYP PAPRPGPLPG PHPSLGSGNE DEDDDEAGGE EDVDDEVFEA SEGARPGSRM PLKSPVPFLP GTSPSADGPD SFSCVFEAIL ESHRAKGTSY TSLASLEALA SPGPTQSPFF TFELPPQPPA PRPDPPAPAP LAPLEPDSGT SSAADGPWTQ RGEEEEAEAR AKLAPGREPP SPCHSEDSLG LGAAPLGSEP PLSQLVSDSD SELDSTERLA LGSTDTLSNG QKADLEAAQR LAKRLYRLDG FRKADVARHL GKNNDFSKLV AGEYLKFFVF TGMTLDQALR VFLKELALMG ETQERERVLA HFSQRYFQCN PEALSSEDGA HTLTCALMLL NTDLHGHNIG KRMTCGDFIG NLEGLNDGGD FPRELLKALY SSIKNEKLQW AIDEEELRRS LSELADPNPK VIKRISGGSG SGSSPFLDLT PEPGAAVYKH GALVRKVHAD PDCRKTPRGK RGWKSFHGIL KGMILYLQKE EYKPGKALSE TELKNAISIH HALATRASDY SKRPHVFYLR TADWRVFLFQ APSLEQMQSW ITRINVVAAM FSAPPFPAAV SSQKKFSRPL LPSAATRLSQ EEQVRTHEAK LKAMASELRE HRAAQLGKKG RGKEAEEQRQ KEAYLEFEKS RYSTYAALLR VKLKAGSEEL DAVEAALAQA GSTEDGLPPS HSSPSLQPKP SSQPRAQRHS SEPRPGAGSG RRKP

          Target Information

          PSD, which called EFA6A, is 63% identical to EFA6B (PSD4) and contains a Sec7 domain adjacent to a PH domain, followed by a C-terminal coiled-coil motif. This gene is mapped to chromosome 10q24. And it encodes a Plekstrin homology and SEC7 domains-containing protein that functions as a guanine nucleotide exchange factor. The encoded protein regulates signal transduction by activating ADP-ribosylation factor 6. Alternative splicing results in multiple transcript variants.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Exchange factor for ADP-ribosylation factor guanine nucleotide factor 6; Exchange factor for ARF6; Exchange factor for ARF6 A; PH and SEC7 domain-containing protein 1; Pleckstrin homology and SEC7 domain-containing protein 1; pleckstrin-Sec7 domains-containing

          View more View less

          Gene Aliases: EFA6; EFA6A; KIAA2011; PSD; PSD1; TYL

          View more View less

          UniProt ID: (Human) A5PKW4

          View more View less

          Entrez Gene ID: (Human) 5662, (Rat) 171381

          View more View less

          Function(s)
          signal transducer activity ARF guanyl-nucleotide exchange factor activity protein binding phospholipid binding guanine nucleotide transmembrane transporter activity
          Process(es)
          signal transduction neuron differentiation regulation of ARF protein signal transduction positive regulation of GTPase activity guanine nucleotide transport guanine transport vesicle-mediated transport
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-f7m9l:80/100.66.129.252:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0