Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Peptide Sequence: MPAHSLVMSS PALPAFLLCS TLLVIKMYVV AIITGQVRLR KKAFANPEDA LRHGGPQYCR SDPDVERCLR AHRNDMETIY PFLFLGFVYS FLGPNPFVAW MHFLVFLVGR VAHTVAYLGK LRAPIRSVTY TLAQLPCASM ALQILWEAAR HL
PTGES is a glutathione-dependent prostaglandin E synthase. The expression of this gene has been shown to be induced by proinflammatory cytokine interleukin 1 beta (IL1B). Its expression can also be induced by tumor suppressor protein TP53, and may be involved in TP53 induced apoptosis.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Glutathione peroxidase PTGES; glutathione S-transferase 1-like 1; Glutathione transferase PTGES; MGST1-L1; MGST1-like 1; Microsomal glutathione S-transferase 1-like 1; Microsomal prostaglandin E synthase 1; microsomal prostaglandin E synthase-1; MPGES-1; p53-induced apoptosis protein 12; p53-induced gene 12 protein; Prostaglandin E synthase; tumor protein p53 inducible protein 12
Gene Aliases: MGST-IV; MGST1-L1; MGST1L1; MPGES; mPGES-1; MPGES1; PGES; PIG12; PP102; PP1294; PTGES; TP53I12
UniProt ID: (Human) O14684
Entrez Gene ID: (Human) 9536
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support