Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • FAK Antibodies

          Abnova

          PTK2 Polyclonal Antibody, MaxPab™

          View all (101) FAK antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite PTK2 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (4)
          PTK2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PTK2 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 4

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PTK2 Antibody (H00005747-D01P) in WB

          Western Blot analysis of PTK2 expression in transfected 293T cell line (H00005747-T01) by PTK2 MaxPab polyclonal antibody.

          Lane 1: PTK2 transfected lysate(76.90 KDa).
          Lane 2: Non-transfected lysate.
          {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PTK2 Antibody in Western Blot (WB)
          PTK2 Antibody in Proximity Ligation Assay (PLA) (PLA)
          PTK2 Antibody in Proximity Ligation Assay (PLA) (PLA)
          PTK2 Antibody in Proximity Ligation Assay (PLA) (PLA)

          Product Details

          H00005747-D01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1-5 µg/mL
          -

          in situ PLA (PLA)

          1:1200
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PTK2 (AAH28733.1, 1 a.a. approximately 680 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          See Label

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Sequence of this protein is as follows: MKYQEVRCLT SFNISVSFPA ETDDYAEIID EEDTYTMPST RDYEIQRERI ELGRCIGEGQ FGDVHQGIYM SPENPALAVA IKTCKNCTSD SVREKFLQEA LTMRQFDHPH IVKLIGVITE NPVWIIMELC TLGELRSFLQ VRKYSLDLAS LILYAYQLST ALAYLESKRF VHRDIAARNV LVSSNDCVKL GDFGLSRYME DSTYYKASKG KLPIKWMAPE SINFRRFTSA SDVWMFGVCM WEILMHGVKP FQGVKNNDVI GRIENGERLP MPPNCPPTLY SLMTKCWAYD PSRRPRFTEL KAQLSTILEE EKAQQEERMR MESRRQATVS WDSGGSDEAP PKPSRPGYPS PRSSEGFYPS PQHMVQTNHY QVSGYPGSHG ITAMAGSIYP GQASLLDQTD SWNHRPQEIA MWQPNVEDST VLDLRGIGQV LPTHLMEERL IRQQQEMEED QRWLEKEERF LKPDVRLSRG SIDREDGSLQ GPIGNQHIYQ PVGKPDPAAP PKKPPRPGAP GHLGSLASLS SPADSYNEGV KLKPQEISPP PTANLDRSND KVYENVTGLV KAVIEMSSKI QPAPPEEYVP MVKEVGLALR TLLATVDETI PLLPASTHRE IEMAQKLLNS DLGELINKMK LAQQYVMTSL QQEYKKQMLT AAHALAVDAK NLLDVIDQAR LKMLGQTRRH

          Target Information

          Focal Adhesion Kinase (FAK) is a 125 kDa non-receptor protein tyrosine kinase that acts as a substrate for Src and is a key element of integrin signaling. FAK plays an important role in cell spreading, differentiation, migration, cell death, and acceleration of the G1 to S phase transition of the cell cycle. FAK has a central catalytic domain and a C-terminal tail that localizes it to focal adhesions, which are sites where cells attach to the extracellular matrix via surface integrin receptors. Increased FAK tyrosine phosphorylation occurs upon integrin engagement with fibronectin. Adhesion of murine NIH3T3 fibroblasts to fibronectin promotes association of the Grb2 adapter protein and c-Src PTK with FAK in vivo, and also results in activation of the ERK2 MAP kinase. In v-Src-transformed NIH3T3, the association of v-Src, Grb2, and Sos with FAK is independent of cell adhesion to fibronectin. In vitro the Grb2 SH2 domain binds directly to tyrosine-phosphorylated FAK, and the binding site has been identified as Tyr925 by site directed mutagenesis. Several transcript variants encoding different isoforms have been found for the FAK gene, but the full-length natures of only three of them have been determined.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: EC 2.7.10.2; FADK 1; FAK-related non-kinase polypeptide; Focal adhesion kinase 1; focal adhesion kinase isoform FAK-Del33; Focal adhesion kinase-related nonkinase; FRNK; p125FAK; pp125 PTK2; pp125FAK; PPP1R71; Protein phosphatase 1 regulatory subunit 71; protein phosphatase 1, regulatory subunit 71; Protein-tyrosine kinase 2; PTK2 protein tyrosine kinase 2

          View more View less

          Gene Aliases: FADK; FAK; FAK1; FRNK; p125FAK; pp125FAK; PPP1R71; PTK2

          View more View less

          UniProt ID: (Human) Q05397

          View more View less

          Entrez Gene ID: (Human) 5747

          View more View less

          Function(s)
          actin binding protein kinase activity protein tyrosine kinase activity non-membrane spanning protein tyrosine kinase activity signal transducer activity Ras guanyl-nucleotide exchange factor activity receptor binding protein binding ATP binding JUN kinase binding protein kinase binding SH2 domain binding
          Process(es)
          MAPK cascade microtubule cytoskeleton organization angiogenesis vasculogenesis neuron migration placenta development regulation of protein phosphorylation positive regulation of protein phosphorylation heart morphogenesis cellular component disassembly involved in execution phase of apoptosis signal complex assembly epidermal growth factor receptor signaling pathway transforming growth factor beta receptor signaling pathway integrin-mediated signaling pathway axon guidance positive regulation of cell proliferation regulation of cell shape embryo development regulation of endothelial cell migration regulation of epithelial cell migration positive regulation of phosphatidylinositol 3-kinase signaling peptidyl-tyrosine phosphorylation central nervous system neuron axonogenesis negative regulation of cell-cell adhesion establishment of cell polarity extracellular matrix organization positive regulation of cell migration regulation of cell adhesion mediated by integrin netrin-activated signaling pathway peptidyl-tyrosine autophosphorylation Fc-gamma receptor signaling pathway involved in phagocytosis establishment of nucleus localization regulation of cell proliferation negative regulation of apoptotic process regulation of GTPase activity endothelial cell migration positive regulation of GTPase activity positive regulation of phosphatidylinositol 3-kinase activity innate immune response regulation of osteoblast differentiation positive regulation of protein kinase activity negative regulation of organ growth protein autophosphorylation vascular endothelial growth factor receptor signaling pathway ephrin receptor signaling pathway cell motility negative regulation of axonogenesis regulation of cytoskeleton organization regulation of focal adhesion assembly positive regulation of protein kinase B signaling negative regulation of synapse assembly growth hormone receptor signaling pathway regulation of substrate adhesion-dependent cell spreading positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process negative regulation of anoikis
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67c77f6dc7-n6nf2:80/100.66.135.186:80.
          git-commit: 545e792480003146e539d624bd2a21b51d198228
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0