Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
The Isotype of this product is composed of an IgG Mixture.
Immunogen sequence: MSGVSEPLSR VKLGTLRRPE GPAEPMVVVP VDVEKEDVRI LKVCFYSNSF NPGKNFKLVK CTVQTEIREI ITSILLSGRI GPNIRLAECY GLRLKHMKSD EIHWLHPQMT VGEVQDKYEC LHVEAEWRYD LQIRYLPEDF MESLKEDRTT LLYFYQQLRN DYMQRYASKV SEGMALQLGC LELRRFFKDM PHNALDKKSN FELLEKEVGL DLFFPKQMQE NLKPKQFRKM IQQTFQQYAS LREEECVMKF FNTLAGFANI DQETYRCELI QGWNITVDLV IGPKGIRQLT SQDAKPTCLA EFKQIRSIRC LPLEEGQAVL QLGIEGAPQA LSIKTSSLAE AENMADLIDG YCRLQGEHQG SLIIHPRKDG EKRNSLPQIP MLNLEARRSH LSESCSIESD IYAEIPDETL RRPGGPQYGI AREDVVLNRI LGEGFFGEVY EGVYTNHKGE KINVAVKTCK KDCTLDNKEK FMSEAVIMKN LDHPHIVKLI GIIEEEPTWI IMELYPYGEL GHYLERNKNS LKVLTLVLYS LQICKAMAYL ESINCVHRDI AVRNILVASP ECVKLGDFGL SRYIEDEDYY KASVTRLPIK WMSPESINFR RFTTASDVWM FAVCMWEILS FGKQPFFWLE NKDVIGVLEK GDRLPKPDLC PPVLYTLMTR CWDYDPSDRP RFTELVCSLS DVYQMEKDIA MEQERNARYR TPKILEPTAF QEPPPKPSRP KYRPPPQTNL LAPKLQFQVP EGLCASSPTL TSPMEYPSPV NSLHTPPLHR HNVFKRHSMR EEDFIQPSSR EEAQQLWEAE KVKMRQILDK QQKQMVEDYQ WLRQEEKSLD PMVYMNDKSP LTPEKEVGYL EFTGPPQKPP RLGAQSIQPT ANLDRTDDLV YLNVMELVRA VLELKNELCQ LPPEGYVVVV KNVGLTLRKL IGSVDDLLPS LPSSSRTEIE GTQKLLNKDL AELINKMRLA QQNAVTSLSE ECKRQMLTAS HTLAMDAKNL LDAVDQAKVL ANLAHPPAE
PTK2B (FAK2) is a cytoplasmic tyrosine kinase involved in calcium-induced regulation of ion channels and activation of the map kinase signaling pathway. PTK2B regulates reorganization of the actin cytoskeleton, cell polarization, cell migration, adhesion, spreading, and bone remodeling. FAK2 plays a role in the regulation of the humoral immune response and is required for normal levels of marginal B-cells in the spleen and normal migration of splenic B-cells. It functions in signaling downstream of integrin and collagen receptors, immune receptors, G-protein coupled receptors, cytokine, chemokine, and growth factor receptors, and mediates responses to cellular stress. Aberrant FAK2 expression may play a role in cancer cell proliferation, migration, and invasion in tumor formation and metastasis. Elevated expression has been seen in gliomas, hepatocellular carcinoma, lung cancer, and breast cancer.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CADTK; CAK beta; CAK-beta; Calcium-dependent tyrosine kinase; Calcium-regulated non-receptor proline-rich tyrosine kinase; Cell adhesion kinase beta; FADK; FADK 2; FAK; FAK1; Focal adhesion kinase 2; FRNK; pp125FAK; Proline-rich tyrosine kinase 2; protein kinase B; Protein-tyrosine kinase 2-beta; PTK2 protein tyrosine kinase 2 beta; PTK2B protein tyrosine kinase 2 beta; RAFTK; Related adhesion focal tyrosine kinase
Gene Aliases: CADTK; CAKB; CAKbeta; FADK2; FAK2; PKB; PTK; PTK2B; PYK2; RAFTK
UniProt ID: (Human) Q14289, (Rat) P70600
Entrez Gene ID: (Human) 2185, (Rat) 50646
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support