Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: MSDAAVDTSS EITTKDLKEK KEVVEEAENG RDAPANGNAN EENGEQEADN EVDEEEEEGG EEEEEEEEGD GEEEDGDEDE EAESATGKRA AEDDEDDDVD TKKQKTDEDD
The thymus gland produces several hormones or hormone-like substances which are derived from a polypeptide precursor containing (in the rat) 113 amino acids and known as prothymosin-alpha. A peptide containing 28 amino acid residues, named thymosin-alpha-1, was originally isolated from calf thymosin fraction 5 and shown to restore various aspects of immune function in several in vitro and invivo test systems. Thymosin-alpha-1 was subsequently isolated from a similar fraction from human thymosin and reported to have the same amino acid sequence as bovine thymosin-alpha-1. In 2003, Jiang et al. identified a pathway that regulates mitochondria-initiated caspase activity. In this pathway, putative human HLA class II-associated protein (PHAP) promoted caspase-9 activation after apoptosome formation, whereas prothymosin-alpha-1 negatively regulated caspase-9 activation by inhibiting apoptosome formation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: gene sequence 28; Prothymosin alpha; prothymosin alpha protein; thymosin; TMSB4; TMSB4X
Gene Aliases: PTMA; TMSA
UniProt ID: (Human) P06454
Entrez Gene ID: (Human) 5757
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support