Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
PTOV1 may activate transcription. Required for nuclear translocation of FLOT1. Promotes cell proliferation.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Activator interaction domain-containing protein 2; granule cell antiserum positive 3; prostate tumor over expressed gene 1; Prostate tumor-overexpressed gene 1 protein; Prostate tumor-overexpressed gene 1 protein homolog; PTOV-1
Gene Aliases: 1110030G05Rik; ACID2; AU041779; Gcap3; PP642; PTOV-1; PTOV1; UNQ6127/PRO20092
UniProt ID: (Human) Q86YD1, (Mouse) Q91VU8, (Rat) Q5U2W6
Entrez Gene ID: (Human) 53635, (Mouse) 84113, (Rat) 292888
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support