Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Sequence of this protein is as follows: MAPGVARGPT PYWRLRLGGA ALLLLLIPVA AAQEPPGAAC SQNTNKTCEE CLKNVSCLWC NTNKACLDYP VTSVLPPASL CKLSSARWGV CWVNFEALII TMSVVGGTLL LGIAICCCCC CRRKRSRKPD RSEEKAMRER EERRIRQEER RAEMKTRHDE IRKKYGLFKE ENPYARFENN
This gene encodes a single-pass type I integral membrane protein, which binds to pituitary tumor-transforming 1 protein (PTTG1), and facilitates translocation of PTTG1 into the nucleus. Coexpression of this protein and PTTG1 induces transcriptional activation of basic fibroblast growth factor. Alternatively spliced transcript variants have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: PBF; Pituitary tumor-transforming gene 1 protein-interacting protein; Pituitary tumor-transforming gene protein-binding factor; PTTG-binding factor
Gene Aliases: C21orf1; C21orf3; PBF; PTTG1IP
UniProt ID: (Human) P53801
Entrez Gene ID: (Human) 754
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support