Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PUM1 Antibodies

          Abnova

          PUM1 Polyclonal Antibody, MaxPab™

          View all (13) PUM1 antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite PUM1 Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          PUM1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PUM1 Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PUM1 Antibody (H00009698-B01P) in WB

          Western Blot analysis of PUM1 expression in transfected 293T cell line by PUM1 MaxPab polyclonal antibody. Lane 1: PUM1 transfected lysate(130.46 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PUM1 Antibody in Western Blot (WB)

          Product Details

          H00009698-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PUM1 (NP_055491, 1 a.a. approximately 1186 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.43 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MSVACVLKRK AVLWQDSFSP HLKHHPQEPA NPNMPVVLTS GTGSQAQPQP AANQALAAGT HSSPVPGSIG VAGRSQDDAM VDYFFQRQHG EQLGGGGSGG GGYNNSKHRW PTGDNIHAEH QVRSMDELNH DFQALALEGR AMGEQLLPGK KFWETDESSK DGPKGIFLGD QWRDSAWGTS DHSVSQPIMV QRRPGQSFHV NSEVNSVLSP RSESGGLGVS MVEYVLSSSP GDSCLRKGGF GPRDADSDEN DKGEKKNKGT FDGDKLGDLK EEGDVMDKTN GLPVQNGIDA DVKDFSRTPG NCQNSANEVD LLGPNQNGSE GLAQLTSTNG AKPVEDFSNM ESQSVPLDPM EHVGMEPLQF DYSGTQVPVD SAAATVGLFD YNSQQQLFQR PNALAVQQLT AAQQQQYALA AAHQPHIGLA PAAFVPNPYI ISAAPPGTDP YTAGLAAAAT LGPAVVPHQY YGVTPWGVYP ASLFQQQAAA AAAATNSANQ QTTPQAQQGQ QQVLRGGASQ RPLTPNQNQQ GQQTDPLVAA AAVNSALAFG QGLAAGMPGY PVLAPAAYYD QTGALVVNAG ARNGLGAPVR LVAPAPVIIS SSAAQAAVAA AAASANGAAG GLAGTTNGPF RPLGTQQPQP QPQQQPNNNL ASSSFYGNNS LNSNSQSSSL FSQGSAQPAN TSLGFGSSSS LGATLGSALG GFGTAVANSN TGSGSRRDSL TGSSDLYKRT SSSLTPIGHS FYNGLSFSSS PGPVGMPLPS QGPGHSQTPP PSLSSHGSSS SLNLGGLTNG SGRYISAAPG AEAKYRSASS ASSLFSPSST LFSSSRLRYG MSDVMPSGRS RLLEDFRNNR YPNLQLREIA GHIMEFSQDQ HGSRFIQLKL ERATPAERQL VFNEILQAAY QLMVDVFGNY VIQKFFEFGS LEQKLALAER IRGHVLSLAL QMYGCRVIQK ALEFIPSDQQ NEMVRELDGH VLKCVKDQNG NHVVQKCIEC VQPQSLQFII DAFKGQVFAL STHPYGCRVI QRILEHCLPD QTLPILEELH QHTEQLVQDQ YGNYVIQHVL EHGRPEDKSK IVAEIRGNVL VLSQHKFASN VVEKCVTHAS RTERAVLIDE VCTMNDGPHS ALYTMMKDQY ANYVVQKMID VAEPGQRKIV MHKIRPHIAT LRKYTYGKHI LAKLEKYYMK NGVDLGPICG PPNGII

          Target Information

          PUM1 (pumilio homolog 1) is a sequence-specific RNA-binding protein that acts as a post-transcriptional repressor by binding the 3'-UTR of mRNA targets. It is a member of the PUF family comprised of evolutionarily conserved RNA-binding proteins. PUM1 binds to an RNA consensus sequence, the Pumilio Response Element (PRE), and 5'-UGUANAUA-3' that is related to the Nanos Response Element (NRE). PUM1 mediates post-transcriptonal repression of transcripts via different mechanisms - acts via direct recruitment of the CCR4-POP2-NOT deadenylase leading to translational inhibition and mRNA degradation. Mutations to the gene can result in spinocerebellar ataxia 47.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: HsPUM; Pumilio homolog 1; pumilio RNA-binding family member 1; pumilio-1

          View more View less

          Gene Aliases: HSPUM; KIAA0099; PUM1; PUMH; PUMH1; PUML1

          View more View less

          UniProt ID: (Human) Q14671

          View more View less

          Entrez Gene ID: (Human) 9698

          View more View less

          Function(s)
          RNA binding mRNA 3'-UTR binding protein binding poly(A) RNA binding
          Process(es)
          spermatogenesis adult locomotory behavior posttranscriptional regulation of gene expression posttranscriptional gene silencing regulation of mRNA stability stem cell differentiation regulation of cell cycle regulation of chromosome segregation regulation of gene silencing by miRNA mRNA destabilization positive regulation of RIG-I signaling pathway positive regulation of gene silencing by miRNA
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0