Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: MADRDSGSEQ GGAALGSGGS LGHPGSGSGS GGGGGGGGGG GGSGGGGGGA PGGLQHETQE LASKRVDIRN KRFYLDVKQN AKGRFLKIAE VGAGGNKSRL TLSMSVAVEF RDYLGDFIEH YAQLGPSQPP DLAQAQDEPR RALKSEFLVR ENRKYYMDLK ENQRGRFLRI RQTVNRGPGL GSTQGQTIAL PAQGLIEFRD ALAKLIDDYG VEEEPAELPE GTSLTVDNKR FFFDVGSNKY GVFMRVSEVK PTYRNSITVP YKVWAKFGHT FCKYSEEMKK IQEKQREKRA ACEQLHQQQQ QQQEETAAAT LLLQGEEEGE ED
This gene product is a sequence-specific, single-stranded DNA-binding protein. It binds preferentially to the single strand of the purine-rich element termed PUR, which is present at origins of replication and in gene flanking regions in a variety of eukaryotes from yeasts through humans. Thus, it is implicated in the control of both DNA replication and transcription. Deletion of this gene has been associated with myelodysplastic syndrome and acute myelogenous leukemia.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Pur alpha; pura {ECO:0000250|UniProtKB:P42669}; purine rich element binding protein A; purine-rich element binding protein A; Purine-rich single-stranded DNA-binding protein alpha; purine-rich single-stranded DNA-binding protein alpha {ECO:0000250|UniProtKB:P42669}; Transcriptional activator protein Pur-alpha; transcriptional activator protein Pur-alpha {ECO:0000250|UniProtKB:P42669}
Gene Aliases: CAGER-1; MRD31; PUR-ALPHA; PUR1; PURA; PURALPHA; ssCRE-BP
UniProt ID: (Human) Q00577, (Mouse) P42669
Entrez Gene ID: (Human) 5813, (Mouse) 19290, (Rat) 307498
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support