Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PXDN Antibodies

          Abnova

          PXDN Polyclonal Antibody, MaxPab™

          View all (5) PXDN antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite PXDN Polyclonal Antibody, MaxPab™

          • Antibody Testing Data (1)
          PXDN Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          PXDN Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          PXDN Antibody (H00007837-B01P) in WB

          Western Blot analysis of PXDN expression in transfected 293T cell line by PXDN MaxPab polyclonal antibody. Lane 1: PXDN transfected lysate(162.69 KDa). Lane 2: Non-transfected lysate. . {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          PXDN Antibody in Western Blot (WB)

          Product Details

          H00007837-B01P

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human

          Host/Isotype

          Mouse / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PXDN (AAI40296.1, 1 a.a. approximately 1479 a.a) full-length human protein.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          PBS, pH 7.4

          Contains

          no preservative

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Product may be used with Western Blot (Transfected lysate).

          The Isotype of this product is composed of an IgG Mixture.

          Immunogen sequence: MAKRSRGPGR RCLLALVLFC AWGTLAVVAQ KPGAGCPSRC LCFRTTVRCM HLLLEAVPAV APQTSILDLR FNRIREIQPG AFRRLRNLNT LLLNNNQIKR IPSGAFEDLE NLKYLYLYKN EIQSIDRQAF KGLASLEQLY LHFNQIETLD PDSFQHLPKL ERLFLHNNRI THLVPGTFNH LESMKRLRLD SNTLHCDCEI LWLADLLKTY AESGNAQAAA ICEYPRRIQG RSVATITPEE LNCERPRITS EPQDADVTSG NTVYFTCRAE GNPKPEIIWL RNNNELSMKT DSRLNLLDDG TLMIQNTQET DQGIYQCMAK NVAGEVKTQE VTLRYFGSPA RPTFVIQPQN TEVLVGESVT LECSATGHPP PRISWTRGDR TPLPVDPRVN ITPSGGLYIQ NVVQGDSGEY ACSATNNIDS VHATAFIIVQ ALPQFTVTPQ DRVVIEGQTV DFQCEAKGNP PPVIAWTKGG SQLSVDRRHL VLSSGTLRIS GVALHDQGQY ECQAVNIIGS QKVVAHLTVQ PRVTPVFASI PSDTTVEVGA NVQLPCSSQG EPEPAITWNK DGVQVTESGK FHISPEGFLT INDVGPADAG RYECVARNTI GSASVSMVLS VNVPDVSRNG DPFVATSIVE AIATVDRAIN STRTHLFDSR PRSPNDLLAL FRYPRDPYTV EQARAGEIFE RTLQLIQEHV QHGLMVDLNG TSYHYNDLVS PQYLNLIANL SGCTAHRRVN NCSDMCFHQK YRTHDGTCNN LQHPMWGASL TAFERLLKSV YENGFNTPRG INPHRLYNGH ALPMPRLVST TLIGTETVTP DEQFTHMLMQ WGQFLDHDLD STVVALSQAR FSDGQHCSNV CSNDPPCFSV MIPPNDSRAR SGARCMFFVR SSPVCGSGMT SLLMNSVYPR EQINQLTSYI DASNVYGSTE HEARSIRDLA SHRGLLRQGI VQRSGKPLLP FATGPPTECM RDENESPIPC FLAGDHRANE QLGLTSMHTL WFREHNRIAT ELLKLNPHWD GDTIYYETRK IVGAEIQHIT YQHWLPKILG EVGMRTLGEY HGYDPGINAG IFNAFATAAF RFGHTLVNPL LYRLDENFQP IAQDHLPLHK AFFSPFRIVN EGGIDPLLRG LFGVAGKMRV PSQLLNTELT ERLFSMAHTV ALDLAAINIQ RGRDHGIPPY HDYRVYCNLS AAHTFEDLKN EIKNPEIREK LKRLYGSTLN IDLFPALVVE DLVPGSRLGP TLMCLLSTQF KRLRDGDRLW YENPGVFSPA QLTQIKQTSL ARILCDNADN ITRVQSDVFR VAEFPHGYGS CDEIPRVDLR VWQDCCEDCR TRGQFNAFSY HFRGRRSLEF SYQEDKPTKK TRPRKIPSVG RQGEHLSNST SAFSTRSDAS GTNDFREFVL EMQKTITDLR TQIKKLESRL STTECVDAGG ESHANNTKWK KDACTICECK DGQVTCFVEA CPPATCAVPV NIPGACCPVC LQKRAEEKP

          Target Information

          Drosophila peroxidasin is an extracellular matrix-associated peroxidase (Horikoshi et al., 1999 [PubMed 10441517]). It is expressed exclusively in hemocytes derived from head mesoderm at a very early stage of differentiation. Peroxidasin exists as a homotrimer with a unique hybrid structure that combines an enzymatically functional peroxidase domain with motifs that are typically found in extracellular matrix-associated proteins. It is a secreted protein that contains a secretory recognition sequence at its N terminus. Peroxidasin catalyzes hydrogen peroxide-driven radioiodination, oxidations, and the formation of dityrosine in vitro. It is also thought to function in extracellular matrix consolidation, phagocytosis, and defense.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: hsPxd01; Melanoma-associated antigen MG50; p53-responsive gene 2 protein; Peroxidasin 1; Peroxidasin homolog; Vascular peroxidase 1

          View more View less

          Gene Aliases: COPOA; D2S448; D2S448E; KIAA0230; MG50; PRG2; PXD01; PXDN; PXN; VPO; VPO1

          View more View less

          UniProt ID: (Human) Q92626

          View more View less

          Entrez Gene ID: (Human) 7837

          View more View less

          Function(s)
          peroxidase activity interleukin-1 receptor antagonist activity extracellular matrix structural constituent heme binding metal ion binding
          Process(es)
          negative regulation of cytokine-mediated signaling pathway immune response response to oxidative stress extracellular matrix organization hydrogen peroxide catabolic process oxidation-reduction process cellular oxidant detoxification
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-6659c69d8f-9267x:80/100.66.134.224:80.
          git-commit: 1827ea18100b76eacea45a9906e00cdefce2d263
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0