Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Abnova
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Product may be used with Western Blot (Transfected lysate).
Immunogen sequence: PDSVPVDSFP PAPSMAVVAV LESTLSCEAC KNGMPTISRL LQMPLFSDVL LTTSEKPQFK IPTKLKEALR IAKECIEKRL IEEQKQIHQH RRLTRAQSHH
PXK is widely expressed in all tissues examined except in heart. It binds to and modulates brain Na,K-ATPase subunits ATP1B1 and ATP1B3 and may thereby participate in the regulation of electrical excitability and synaptic transmission. However, the protein kinase domain is predicted to be catalytically inactive.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Modulator of Na,K-ATPase; modulator of Na,K-ATPase long form; MONaKA; PX domain-containing protein kinase-like protein; PX ser/thr kinase v2; PX serine/threonine kinase; PXK
Gene Aliases: MONaKA; PXK
UniProt ID: (Human) Q7Z7A4
Entrez Gene ID: (Human) 54899
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support