Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
Immunogen sequence: IVMARGRHVG SSETKLLQHL LEACGRCEEV PEAYVDIHTG LSGSGVAFVC AFSEALAEGA VKMGMPSSLA HRIAAQTLLG TAKMLLHEGQ HPAQLRSDVC TPGGTTIYGL HALEQGGLRA ATMSAVEAAT CRAKELSRK
PYCRL encodes a protein that belongs to the pyrroline-5-carboxylate reductase family of enzymes. Members of this family catalyze the final step in proline biosynthesis, converting pyrroline-5-carboxylate to proline. Glutamate and ornithine are precursors in the synthesis of proline. The protein encoded by this gene is a cytoplasmic enzyme involved in the biosynthesis of proline from ornithine.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: P5C reductase 3; P5CR 3; Pyrroline-5-carboxylate reductase 3; Pyrroline-5-carboxylate reductase-like protein
Gene Aliases: PYCR3; PYCRL
UniProt ID: (Human) Q53H96
Entrez Gene ID: (Human) 65263
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support