Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 2
This gene encodes a member of the peptidyl arginine deiminase family of enzymes, which catalyze the post-translational deimination of proteins by converting arginine residues into citrullines in the presence of calcium ions. The family members have distinct substrate specificities and tissue-specific expression patterns. The type I enzyme is involved in the late stages of epidermal differentiation, where it deiminates filaggrin and keratin K1, which maintains hydration of the stratum corneum, and hence the cutaneous barrier function. This enzyme may also play a role in hair follicle formation. This gene exists in a cluster with four other paralogous genes.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hPAD-colony 10; Pad type 1; PAD type I; PADI5; PDI2; PDI3; PDI4; peptidyl arginine deiminase, type I; Peptidylarginine deiminase I; Protein-arginine deiminase type I; PROTEIN-ARGININE DEIMINASE TYPE I (PEPTIDYLARGININE DEIMINASE I); Protein-arginine deiminase type-1
Gene Aliases: AV236283; HPAD10; PAD1; PADI1; PDI; PDI1
UniProt ID: (Human) Q9ULC6, (Mouse) Q9Z185
Entrez Gene ID: (Human) 29943, (Mouse) 18599
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support