Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: LELLGRERD RVQVERDGLA EDLAALKQRL EEETRKREDA EHNLVLFRKD VDDATLSRLE LERKIESLMD EIEFLKKLHE EELRDLQVSV ESQQVQQVEV EATVKPELTA ALRDIRAQYE SIAAKNLQEA EEWYKSKYAD LSDAANRNHE ALRQAKQEMN ESRRQIQSLT CEVDGLRGTN EALLRQLREL EEQFALEAGG YQAGAARLEE ELRQLKEEMA RHLREYQELL NVKMALDIEI ATYRKLLEGE ESRISVPVHS FASLNIKTTV PEVEPPQDSH SRKTVLIKTI ETRNGEVVTE SQKEQRSELD KSSAHSY (155-470 aa encoded by BC032703 )
Peripherin is a 57kD type III intermediate filament that is a specific marker for peripheral neurons, including enteric ganglion cells. Peripherin is expressed in the developing peripheral nervous system and is highly enriched in neuronal derivatives of the neural crest. Peripherin offers an advantage over other neural markers, such as S100 and NSE in that it does not stain chrmaffin cell types.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Neurofilament 4; neurofilament 4 (57kD); Peripherin; peripherin 1
Gene Aliases: NEF4; Perf; PRPH; PRPH1
UniProt ID: (Human) P41219
Entrez Gene ID: (Human) 5630, (Rat) 24688
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support