Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CD61 (Integrin beta 3) Antibodies

          Invitrogen

          Phospho-CD61 (Tyr785) Polyclonal Antibody

          View all (100) CD61 (Integrin beta 3) antibodies
          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-CD61 (Tyr785) Polyclonal Antibody

          • Antibody Testing Data (1)
          Phospho-CD61 (Tyr785) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-CD61 (Tyr785) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-CD61 (Tyr785) Antibody (PA5-99351) in WB

          Western blot analysis of Phospho-CD61 (Tyr785) in insulin treated 293 cell lysate (left lane: treated with the antigen-specific peptide). Samples were incubated with Phospho-CD61 (Tyr785) polyclonal antibody (Product # PA5-99351). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-CD61 (Tyr785) Antibody in Western Blot (WB)

          Product Details

          PA5-99351

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human ITGB3(Accession P05106), corresponding to amino acid residues around phosphorylated Tyr785.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          sequential chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2818284

          Product Specific Information

          Antibody detects endogenous levels of Integrin beta3 only when phosphorylated at Tyrosine 785.

          Target Information

          CD61 (GPIIIa, ITGB3) is a glycoprotein found on megakaryocytes, platelets and their precursors. CD61 antigen plays a role in platelet aggregation and also as a receptor for fibrinogen, fibronectin, von Willebrand factor and vitronectrin. CD61 is a 105 kDa glycoprotein that associates with either the alpha 11b integrin (CD41) or the alpha V integrin (CD51) at the cell surface. CD61 is expressed on platelets and megakaryocytes in association with CD41, and on endothelial cells, monocytes, platelets and osteoclasts in association with CD51. CD61 is a receptor for fibrinogen, fibronectin, Von Willebrand Factor, vitronectin and thrombospondin. The CD61 protein product is composed of an alpha chain and a beta chain. A given chain may combine with multiple partners resulting in different integrins. CD61 is found along with the alpha IIb chain in platelets. Like other integrins, CD61 is known to participate in cell adhesion as well as cell-surface mediated signaling. Diseases associated with CD61 dysfunction include Glanzmann Thrombasthenia and Platelet type-16 Bleeding Disorder.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: antigen CD61; CD61; GPIIIa; integrin beta 3; integrin beta 3 (Cd61); Integrin beta-3; integrin, beta 3 (platelet glycoprotein IIIa, antigen CD61); platelet gpIIIa; Platelet membrane glycoprotein IIIa

          View more View less

          Gene Aliases: BDPLT16; BDPLT2; CD61; GP3A; GPIIIa; GT; INGRB3; ITGB3

          View more View less

          UniProt ID: (Human) P05106, (Rat) Q8R2H2, (Mouse) O54890

          View more View less

          Entrez Gene ID: (Human) 3690, (Rat) 29302, (Mouse) 16416

          View more View less

          Function(s)
          virus receptor activity fibronectin binding protease binding protein disulfide isomerase activity platelet-derived growth factor receptor binding protein binding coreceptor activity fibroblast growth factor binding enzyme binding C-X3-C chemokine binding insulin-like growth factor I binding neuregulin binding identical protein binding vascular endothelial growth factor receptor 2 binding cell adhesion molecule binding extracellular matrix binding integrin binding peptide binding fibrinogen binding receptor activity
          Process(es)
          positive regulation of protein phosphorylation positive regulation of endothelial cell proliferation platelet degranulation cell-substrate junction assembly cell adhesion cell-matrix adhesion integrin-mediated signaling pathway blood coagulation positive regulation of endothelial cell migration negative regulation of macrophage derived foam cell differentiation negative regulation of lipid storage smooth muscle cell migration cell growth cell migration platelet activation extracellular matrix organization positive regulation of vascular endothelial growth factor receptor signaling pathway cell-substrate adhesion activation of protein kinase activity negative regulation of lipid transport heterotypic cell-cell adhesion substrate adhesion-dependent cell spreading tube development apolipoprotein A-I-mediated signaling pathway wound healing regulation of bone resorption negative regulation of low-density lipoprotein particle receptor biosynthetic process viral entry into host cell vascular endothelial growth factor receptor signaling pathway mesodermal cell differentiation positive regulation of peptidyl-tyrosine phosphorylation negative regulation of lipoprotein metabolic process leukocyte migration negative chemotaxis angiogenesis involved in wound healing platelet aggregation positive regulation of leukocyte migration protein folding cellular calcium ion homeostasis embryo implantation regulation of G-protein coupled receptor protein signaling pathway response to radiation positive regulation of fibroblast migration response to activity positive regulation of smooth muscle cell migration regulation of cell migration positive regulation of cell migration positive regulation of cell adhesion mediated by integrin positive regulation of osteoblast proliferation cellular response to drug response to platelet-derived growth factor positive regulation of MAPK cascade positive regulation of angiogenesis positive regulation of bone resorption positive regulation of smooth muscle cell proliferation regulation of release of sequestered calcium ion into cytosol negative regulation of cell death positive regulation of ERK1 and ERK2 cascade cellular response to mechanical stimulus positive regulation of glomerular mesangial cell proliferation positive regulation of adenylate cyclase-inhibiting opioid receptor signaling pathway cellular response to insulin-like growth factor stimulus positive regulation of osteoclast differentiation cell projection morphogenesis
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0