Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of Cdk1/2 only when phosphorylated at Thr14.
Voltage dependent anion channel required for acidification and functions of the Golgi apparatus that may function in counter-ion conductance.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: cdc2-related protein kinase; CDK1; cell cycle controller CDC2; cell cycle p34 CDC2 kinase protein; Cell division control protein 2 homolog; cell division cycle 2 homolog A; cell division cycle 2, G1 to S and G2 to M; Cell division cycle control protein 2; cell division cycle control protein 2a; Cell division protein kinase 1; Cell division protein kinase 2; cyclin dependent kinase 2-alpha; Cyclin-dependent kinase 1; Cyclin-dependent kinase 2; p33 protein kinase; p34 protein kinase
Gene Aliases:
A630093N05Rik; CDC2; CDC28A; Cdc2a; CDK1; CDK2; CDKN1; CDKN2; p33(CDK2); p34
UniProt ID: (Human) P06493, (Human) P24941, (Rat) P39951, (Mouse) P11440, (Mouse) P97377
Entrez Gene ID: (Human) 983, (Human) 1017, (Rat) 54237, (Mouse) 12534, (Rat) 362817, (Mouse) 12566
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support