Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 4
Antibody detects endogenous levels of Calpain 2 only when phosphorylated at Ser50.
Mu-Calpain, also known as Calpain-I, and m-calpain, also known as Calpain-II, are intracellular, calcium-dependent cysteine proteases. Mu-calpain has a micromolar sensitivity (thus the mu-) as compared to the millimolar calcium sensitivity of m-calpain.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 80 kDa M-calpain subunit; Calcium-activated neutral proteinase 2; CALP80; calpain 2, (m/II) large subunit; calpain 2, large [catalytic] subunit; calpain 2, large subunit; Calpain large polypeptide L2; Calpain M-type; calpain, large polypeptide L2; Calpain-2 catalytic subunit; Calpain-2 large subunit; CANP 2; cysteine proteinase; M-calpain; m-calpain 80 kDa large subunit; m-calpain large subunit; Millimolar-calpain
Gene Aliases: AI326419; CANP2; CANPL2; CANPml; Capa-2; Capa2; CAPN2; m-calpain; m-calpin; mCANP
UniProt ID: (Human) P17655, (Mouse) O08529, (Rat) Q07009
Entrez Gene ID: (Human) 824, (Mouse) 12334, (Rat) 29154
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support