Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Caveolin 1 Antibodies

          Invitrogen

          Phospho-Caveolin 1 (Tyr14) Polyclonal Antibody

          View all (47) Caveolin 1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-Caveolin 1 (Tyr14) Polyclonal Antibody

          • Antibody Testing Data (1)
          Phospho-Caveolin 1 (Tyr14) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-Caveolin 1 (Tyr14) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-Caveolin 1 (Tyr14) Antibody (PA5-104957) in WB

          Western blot analysis of Phospho-Caveolin 1 (Tyr14) in mouse liver/rat kidney (absence (-) or presence (+) of non-phospho and phospho peptide). Samples were incubated with Phospho-Caveolin 1 (Tyr14) polyclonal antibody (Product # PA5-104957). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-Caveolin 1 (Tyr14) Antibody in Western Blot (WB)

          Product Details

          PA5-104957

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:500
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human CAV1(Accession Q03135), corresponding to amino acid residues around phosphorylated Tyr14.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          sequential chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2816430

          Product Specific Information

          Antibody detects endogenous levels of Caveolin 1 only when phosphorylated at Tyrosine 14.

          Target Information

          Caveolin-1 or CAV1 is a scaffolding proteins within caveolar membranes. It interacts directly with G-protein alpha subunits and can functionally regulate their activity. Caveolin-1 is also involved in the co-stimulatory signal essential for T-cell receptor (TCR) mediated T-cell activation. It can bind with DPP4 and induces T-cell proliferation and NF-kappa-B activation in a T-cell receptor/CD3-dependent manner. Mutations affecting the CAV1 gene can result in congenital generalized lipodystrophy 3, pulmonary hypertension primary 3, or partial lipodystrophy/congenital cataracts, and neurodegeneration syndrome.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: caveolae; caveolin 1, caveolae protein, 22kDa; caveolin caveolae protein 22 kDa; caveolin, caveolae protein 1; caveolin, caveolae protein, 22 kDa; Caveolin-1; cell growth-inhibiting protein 32; HGNC:1527; OTTHUMP00000196475

          View more View less

          Gene Aliases: BSCL3; CAV; Cav-1; CAV1; CGL3; LCCNS; MSTP085; PPH3; VIP21

          View more View less

          UniProt ID: (Human) Q03135, (Mouse) P49817

          View more View less

          Entrez Gene ID: (Human) 857, (Mouse) 12389, (Rat) 25404

          View more View less

          Function(s)
          RNA polymerase I transcription factor binding receptor binding patched binding structural molecule activity protein binding cholesterol binding peptidase activator activity enzyme binding protein kinase binding syntaxin binding protein binding, bridging protein complex scaffold identical protein binding ion channel binding protein heterodimerization activity Rac GTPase binding nitric-oxide synthase binding ATPase binding inward rectifier potassium channel inhibitor activity MDM2/MDM4 family protein binding SNARE binding kinase binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter MAPK cascade inactivation of MAPK activity angiogenesis vasculogenesis response to hypoxia negative regulation of endothelial cell proliferation positive regulation of endothelial cell proliferation negative regulation of cytokine-mediated signaling pathway regulation of the force of heart contraction response to ischemia regulation of the force of heart contraction by chemical signal triglyceride metabolic process calcium ion transport cellular calcium ion homeostasis endocytosis regulation of smooth muscle contraction skeletal muscle tissue development lactation protein localization negative regulation of cell proliferation response to mechanical stimulus positive regulation of signal transduction negative regulation of signal transduction positive regulation of calcium ion transport into cytosol posttranscriptional regulation of gene expression positive regulation of gene expression negative regulation of muscle cell apoptotic process positive regulation of peptidase activity receptor-mediated endocytosis of virus by host cell regulation of fatty acid metabolic process lipid storage regulation of blood coagulation negative regulation of BMP signaling pathway negative regulation of epithelial cell differentiation mammary gland development positive regulation of microtubule polymerization T cell costimulation negative regulation of protein ubiquitination positive regulation of protein ubiquitination negative regulation of protein binding positive regulation of protein binding maintenance of protein location in cell response to progesterone negative regulation of peptidyl-serine phosphorylation positive regulation of peptidyl-serine phosphorylation cholesterol efflux nitric oxide homeostasis positive regulation of toll-like receptor 3 signaling pathway wound healing vasoconstriction negative regulation of tyrosine phosphorylation of Stat5 protein cholesterol homeostasis positive regulation of catalytic activity negative regulation of MAP kinase activity negative regulation of MAPK cascade response to estrogen negative regulation of nitric oxide biosynthetic process negative regulation of neuron differentiation positive regulation of endocytosis positive regulation of vasoconstriction negative regulation of JAK-STAT cascade microtubule polymerization negative regulation of pinocytosis negative regulation of smooth muscle cell proliferation negative regulation of nitric-oxide synthase activity protein oligomerization protein homooligomerization regulation of cytosolic calcium ion concentration response to calcium ion membrane depolarization regulation of peptidase activity calcium ion homeostasis mammary gland involution positive regulation of cell adhesion molecule production negative regulation of protein tyrosine kinase activity negative regulation of ERK1 and ERK2 cascade caveola assembly cellular response to mechanical stimulus cellular response to exogenous dsRNA cellular response to peptide hormone stimulus cellular response to hyperoxia cellular response to transforming growth factor beta stimulus caveolin-mediated endocytosis regulation of heart rate by cardiac conduction angiotensin-activated signaling pathway involved in heart process negative regulation of canonical Wnt signaling pathway positive regulation of canonical Wnt signaling pathway cellular senescence apoptotic signaling pathway regulation of membrane repolarization during action potential regulation of ventricular cardiac muscle cell action potential negative regulation of peptidyl-tyrosine autophosphorylation negative regulation of potassium ion transmembrane transport regulation of cell communication by electrical coupling involved in cardiac conduction regulation of inward rectifier potassium channel activity positive regulation of gap junction assembly receptor internalization involved in canonical Wnt signaling pathway negative regulation of anoikis positive regulation of extrinsic apoptotic signaling pathway positive regulation of intrinsic apoptotic signaling pathway negative regulation of cation channel activity response to nutrient response to bacterium response to gamma radiation vesicle organization cholesterol transport negative regulation of transforming growth factor beta receptor signaling pathway organ regeneration receptor internalization response to drug protein localization to plasma membrane raft leukocyte migration regulation of nitric-oxide synthase activity response to glucocorticoid negative regulation of necroptotic process regulation of cardiac muscle cell action potential involved in regulation of contraction regulation of ruffle assembly positive regulation of ER-associated ubiquitin-dependent protein catabolic process protein localization to basolateral plasma membrane beta-catenin destruction complex disassembly regulation of entry of bacterium into host cell cellular response to starvation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0