Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • CDC37 Antibodies

          Invitrogen

          Phospho-Cdc37 (Ser13) Recombinant Rabbit Monoclonal Antibody (5P8B2)

          View all (28) CDC37 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-Cdc37 (Ser13) Recombinant Rabbit Monoclonal Antibody (5P8B2)

          • Antibody Testing Data (2)
          Phospho-Cdc37 (Ser13) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-Cdc37 (Ser13) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-Cdc37 (Ser13) Antibody (MA5-35890) in WB

          Western blot analysis of Phospho-Cdc37 (Ser13) in extracts of various cell lines. Samples were incubated with Phospho-Cdc37 (Ser13) Monoclonal antibody (Product # MA5-35890) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) secondary antibody at a dilution of 1:10,000. Both 293T cells and NIH/3T3 cells were treated by CIP (20 µL/400 µL) at 37°C for 1 hour. Lysates/proteins: 25 µg per l... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-Cdc37 (Ser13) Antibody in Western Blot (WB)
          Phospho-Cdc37 (Ser13) Antibody in Western Blot (WB)

          Product Details

          MA5-35890

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Expression System

          HEK293 cells

          Class

          Recombinant Monoclonal

          Type

          Antibody

          Clone

          5P8B2

          Immunogen

          A synthetic phosphorylated peptide around S13 of human CDC37 (Q16543)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.09 mg/mL

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol, 0.05% BSA

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2849788

          Product Specific Information

          Immunogen sequence: EVSDD

          Target Information

          In S. cerevisiae, Cdc37 has been shown to be an important regulator of cell division cycle through its action on cyclin-dependent kinases (CDK's). Cdc37 has also been associated with signaling pathways and kinases unrelated to cell cycle control, including v-Src, casein kinase II, MPS1 kinase and the sevenless receptor tyrosine kinase. Interestingly, HSP90 has also been shown to be associated with many of the same cellular targets as Cdc37. It has now been shown that Cdc37 is the same protein previously described as p50 which has been found to be associated with many of the same proteins as HSP90. Among others, Cdc37/p50 has been found complexed with Src-family kinases (Fyn, Yes, Fes, Lck & Fgr), Raf and pp60v-src. More recently, Cdc37 has been shown to be a chaperone independently and in concert with HSP90. In fact, Cdc37 is necessary for the binding of HSP90 to Cdk4.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: CDC 37; CDC37 (cell division cycle 37 S. cerevisiae homolog); CDC37 (cell division cycle 37, S. cerevisiae, homolog); CDC37 cell division cycle 37 homolog; Cdc37 homolog; CDC37 protein; cell division cycle 37 homolog; Hsp90 chaperone protein kinase-targeting subunit; Hsp90 co-chaperone Cdc37; p50Cdc37

          View more View less

          Gene Aliases: CDC37; CDC37A; p50; P50CDC37

          View more View less

          UniProt ID: (Human) Q16543, (Rat) Q63692, (Mouse) Q61081

          View more View less

          Entrez Gene ID: (Human) 11140, (Rat) 114562, (Mouse) 12539

          View more View less

          Function(s)
          protein tyrosine kinase activity protein binding protein kinase regulator activity kinase binding protein kinase binding heat shock protein binding unfolded protein binding chaperone binding Hsp90 protein binding protein C-terminus binding mitogen-activated protein kinase kinase kinase binding protein kinase B binding
          Process(es)
          protein folding protein stabilization regulation of cell cycle regulation of interferon-gamma-mediated signaling pathway regulation of type I interferon-mediated signaling pathway regulation of cyclin-dependent protein serine/threonine kinase activity protein targeting posttranscriptional regulation of gene expression peptidyl-tyrosine phosphorylation ERBB2 signaling pathway mitophagy in response to mitochondrial depolarization regulation of protein kinase activity
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0