Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • DOK1/DOK2 Antibodies

          Invitrogen

          Phospho-DOK1/DOK2 (Tyr362, Tyr337) Polyclonal Antibody

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-DOK1/DOK2 (Tyr362, Tyr337) Polyclonal Antibody

          • Antibody Testing Data (1)
          Phospho-DOK1/DOK2 (Tyr362, Tyr337) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-DOK1/DOK2 (Tyr362, Tyr337) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-DOK1/DOK2 (Tyr362, Tyr337) Antibody (PA5-143681) in WB

          Western blot of Phospho-DOK1/DOK2 (Tyr362, Tyr337) in Jurkat cells stimulated with calyculin A (100 nM, 30 min.) (lanes 1-6) followed by lambda phosphatase (lanes 2 & 6) or alkaline phosphatase (lane 4) treatment. The blots were probed with anti-Dok1 (lanes 1 & 2), Phospho-DOK1/DOK2 (Tyr362, Tyr337) polyclonal antibody (Product # PA5-143681) (lanes 3 & 4), and anti-Dok1 (Ser450) (lanes 5 & 6). {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-DOK1/DOK2 (Tyr362, Tyr337) Antibody in Western Blot (WB)

          Product Details

          PA5-143681

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Phospho-Dok1 (Tyr-362) synthetic peptide (coupled to KLH) corresponds to amino acids surrounding tyrosine 362 in human Dok1. This sequence is conserved in Dok1 from rat and mouse (Tyr-361), and has high homology to Dok2 (Tyr-337). The site is not conserved in other Dok family members.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA, 50% glycerol

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2942909

          Product Specific Information

          This antibody was cross-adsorbed to a phosphotyrosine peptide before affinity purification using phospho-Dok1 (Tyr-362) peptide. The purified antibody detects a band at 62 kDa* corresponding to Dok1 in western blots of human Jurkat cells, but does not detect this band after alkaline phosphatase treatment. Similar to Dok1 (Ser-450) antibody, this antibody also detects an 80 kDa band that has not been identified.

          Target Information

          Doks are a family of adaptor proteins that include six Dok proteins (Dok1 to Dok6), which have an N-terminal pleckstrin homology domain, a central phosphotyrosine binding domain, and a C-terminal region containing multiple tyrosine residues. When phosphorylated, these tyrosines can serve as docking sites for SH2 domain-containing proteins. Dok1 (p62dok) has been shown to bind Ras-GAP, Nck, and Csk. Several tyrosine phosphorylation sites have been identified for Dok1. One site, Tyr-362 (Tyr-361 mouse), is phosphorylated by c-Abl, is required for Nck binding, and may be critical for filopodia formation during fibroblast spreading on fibronectin. Alternatively, Dok1 activity is also regulated by serine phosphorylation. IkappaB Kinase beta phosphorylates several serine sites including Ser-450 in vitro, and TNFalpha, IL-1, and radiation treatment lead to phosphorylation of Ser-443, Ser-446, and Ser-450 in vivo. Phosphorylation of these serine sites may be required for Dok-mediated inhibition of MAPK signaling and stimulation of cell motility.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Docking protein 1; docking protein 1, 62kDa (downstream of tyrosine kinase 1); Downstream of tyrosine kinase 1; downstream of tyrosine kinase-1; p62(dok); p62DOK; pp62

          View more View less

          Gene Aliases: AW557123; Dok; Dok-1; DOK1; P62DOK; pp62

          View more View less

          UniProt ID: (Human) Q99704, (Rat) Q4QQV2, (Mouse) P97465

          View more View less

          Entrez Gene ID: (Human) 1796, (Rat) 312477, (Mouse) 13448

          View more View less

          Function(s)
          receptor signaling protein activity insulin receptor binding protein binding
          Process(es)
          signal transduction cell surface receptor signaling pathway transmembrane receptor protein tyrosine kinase signaling pathway Ras protein signal transduction positive regulation of epidermal growth factor receptor signaling pathway intracellular signal transduction MAPK cascade
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-7fd48b9cb8-69xq6:80/100.66.134.130:80.
          git-commit: fe0b5dd8e7a67b57f27a6e2d26c18f8954d91d59
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.30.1-2025.05.22-1.0