Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • p300 Antibodies

          Bioss

          Phospho-Ep300 (Ser1834) Polyclonal Antibody

          View all (24) p300 antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-Ep300 (Ser1834) Polyclonal Antibody

          • Antibody Testing Data (2)
          Phospho-Ep300 (Ser1834) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.
          Phospho-Ep300 (Ser1834) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-Ep300 (Ser1834) Antibody (BS-5339R) in IHC (P)

          Formalin-fixed and paraffin embedded human colon carcinoma labeled with Rabbit Anti-Ep300 (Ser1834) Polyclonal Antibody, Unconjugated (bs-5339R) at 1:200 followed by conjugation to the secondary antibody and DAB staining. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-Ep300 (Ser1834) Antibody in Immunohistochemistry (Paraffin) (IHC (P))
          Phospho-Ep300 (Ser1834) Antibody in Western Blot (WB)

          Product Details

          BS-5339R

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          1:200
          -

          Immunohistochemistry (Frozen) (IHC (F))

          1:100-1:500
          -

          Immunocytochemistry (ICC/IF)

          1:50-1:200
          -

          ELISA (ELISA)

          1:500-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat, Zebrafish

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          KLH conjugated synthetic phosphopeptide derived from human Ep300 around the phosphorylation site of Ser1834.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Protein A

          Storage buffer

          0.01M TBS, pH 7.4, with 50% glycerol, 1% BSA

          Contains

          0.02% ProClin 300

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Target Information

          EP300 encodes the adenovirus E1A-associated cellular p300 transcriptional co-activator protein. p300 is related by sequence to CPB (CREB-binding protein [CREB: cyclic-AMP responsive element binding protein]), and like CPB can stimulate transcription through activation of CREB. This EP300 activity is specifically inhibited by the adenovirus oncoprotein E1A. EP300 has also been identified as a co-activator of HIF1A (hypoxia-inducible factor 1 alpha), and thus plays a role in the stimulation of hypoxia-induced genes such as VEGF.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: E1A-associated protein p300; E1A-binding protein, 300kD; Histone acetyltransferase p300; Histone butyryltransferase p300; Histone crotonyltransferase p300; p300 HAT; Protein 2-hydroxyisobutyryltransferase p300; Protein lactyltransferas p300; Protein propionyltransferase p300

          View more View less

          Gene Aliases: A430090G16; A730011L11; EP300; KAT3B; P300; p300 HAT; RSTS2

          View more View less

          UniProt ID: (Human) Q09472, (Mouse) B2RWS6

          View more View less

          Entrez Gene ID: (Human) 2033, (Mouse) 328572, (Rat) 170915

          View more View less

          Function(s)
          RNA polymerase II core promoter proximal region sequence-specific DNA binding RNA polymerase II core promoter sequence-specific DNA binding core promoter binding RNA polymerase II activating transcription factor binding transcriptional activator activity, RNA polymerase II transcription regulatory region sequence-specific binding p53 binding DNA binding chromatin binding damaged DNA binding transcription coactivator activity histone acetyltransferase activity lysine N-acetyltransferase activity, acting on acetyl phosphate as donor protein binding beta-catenin binding protein C-terminus binding transcription factor binding zinc ion binding acetyltransferase activity transferase activity, transferring acyl groups chromatin DNA binding protein complex binding activating transcription factor binding peptide N-acetyltransferase activity nuclear hormone receptor binding glucocorticoid receptor binding peroxisome proliferator activated receptor binding bHLH transcription factor binding SMAD binding androgen receptor binding mitogen-activated protein kinase binding NF-kappaB binding pre-mRNA intronic binding protein antigen binding RNA polymerase II transcription factor binding core promoter proximal region DNA binding transcription factor activity, sequence-specific DNA binding transcription cofactor activity antigen binding transferase activity protein kinase binding metal ion binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter response to hypoxia somitogenesis liver development positive regulation of protein phosphorylation stimulatory C-type lectin receptor signaling pathway transcription-coupled nucleotide-excision repair regulation of transcription, DNA-templated transcription from RNA polymerase II promoter protein acetylation internal protein amino acid acetylation apoptotic process DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest positive regulation of transcription from RNA polymerase II promoter involved in unfolded protein response Notch signaling pathway nervous system development heart development skeletal muscle tissue development memory circadian rhythm organ morphogenesis regulation of autophagy positive regulation of glycoprotein biosynthetic process macrophage derived foam cell differentiation positive regulation of cell death positive regulation of muscle atrophy viral process N-terminal peptidyl-lysine acetylation internal peptidyl-lysine acetylation B cell differentiation platelet formation lung development response to cobalt ion positive regulation of protein binding positive regulation of type I interferon production positive regulation of collagen biosynthetic process positive regulation of protein import into nucleus, translocation response to tumor necrosis factor cellular response to UV positive regulation of histone acetylation cellular response to drug megakaryocyte development cellular response to trichostatin A intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator negative regulation of cysteine-type endopeptidase activity involved in apoptotic process positive regulation of DNA binding protein kinase B signaling response to estrogen positive regulation by host of viral transcription histone H3 acetylation histone H4 acetylation histone H2B acetylation fat cell differentiation response to ethanol positive regulation of translation positive regulation of axon extension positive regulation of cell size positive regulation of gene expression, epigenetic positive regulation of proteolysis positive regulation of transcription from RNA polymerase II promoter digestive tract development positive regulation of protein secretion protein stabilization positive regulation of sequence-specific DNA binding transcription factor activity response to calcium ion regulation of cell cycle regulation of angiotensin metabolic process positive regulation of sarcomere organization regulation of androgen receptor signaling pathway regulation of transcription from RNA polymerase II promoter in response to hypoxia protein-DNA complex assembly cellular response to hydrogen peroxide response to fatty acid cellular response to retinoic acid cellular response to cAMP cellular response to glucose stimulus cellular response to mineralocorticoid stimulus cellular response to dexamethasone stimulus regulation of tubulin deacetylation regulation of cellular response to heat regulation of signal transduction by p53 class mediator beta-catenin-TCF complex assembly cellular response to nerve growth factor stimulus negative regulation of miRNA metabolic process transcription, DNA-templated cell cycle response to glucose positive regulation of gene expression histone acetylation cell differentiation positive regulation of cell growth negative regulation of cellular metabolic process positive regulation of cellular metabolic process response to drug positive regulation of transcription, DNA-templated rhythmic process cartilage development negative regulation of cell death response to dexamethasone positive regulation of protein acetylation regulation of nucleic acid-templated transcription response to organic cyclic compound response to retinoic acid response to hydrogen peroxide cellular response to antibiotic cellular response to organic cyclic compound
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67c77f6dc7-n6nf2:80/100.66.135.186:80.
          git-commit: 545e792480003146e539d624bd2a21b51d198228
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0