Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
FIGURE: 1 / 4
Antibody detects endogenous levels of EFNB1 only when phosphorylated at Tyr313/317.
Cell surface transmembrane ligand for Eph receptors, a family of receptor tyrosine kinases which are crucial for migration, repulsion and adhesion during neuronal, vascular and epithelial development. Binds promiscuously Eph receptors residing on adjacent cells, leading to contact-dependent bidirectional signaling into neighboring cells. The signaling pathway downstream of the receptor is referred to as forward signaling while the signaling pathway downstream of the ephrin ligand is referred to as reverse signaling. Binds to receptor tyrosine kinase including EPHA4, EPHA3 and EPHB4. Together with EPHB4 plays a central role in heart morphogenesis and angiogenesis through regulation of cell adhesion and cell migration. EPHB4-mediated forward signaling controls cellular repulsion and segregation from EFNB2-expressing cells. May play a role in constraining the orientation of longitudinally projecting axons.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: CEK5 receptor ligand; CEK5-L; CEL5-L; EFL-3; EFL6; EFNB3; ELK ligand; ELK-L; EPH-related receptor tyrosine kinase ligand 2; Ephrin B3; Ephrin-B1; Ephrin-B1 precursor; EphrinB1; EPLG8; kinase ephrin-B1; LERK-2; LERK8; Stimulated by retinoic acid gene 1 protein; STRA1 protein
Gene Aliases: Cek5-L; CFND; CFNS; EFB1; EFL-3; EFL3; EFNB1; Elk-L; Epl2; EPLG2; LERK-2; LERK2; Stra1
UniProt ID: (Human) P98172, (Mouse) P52795
Entrez Gene ID: (Human) 1947, (Rat) 25186, (Mouse) 13641
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support