Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
This antibody is predicted to react with bovine, canine, chicken, finch, human, mouse, non-human primate, Xenopus and zebrafish based on 100% sequence homology.
This antibody is specific for the ~50 kDa Gap-43 protein phosphorylated at Ser41 in Western blots of rat brain lysate. In some tissues the antibody also recognizes a higher molecular weight protein, that is also recognized by the Pan Gap-43 antibody, that may be a Gap-43 aggregate or oligomer. Immunolabeling blocked by the phosphopeptide used as the antigen but not by the corresponding dephosphopeptide.
GAP43 (Neuromodulin) is associated with nerve growth. GAP43 is expressed by developing and regenerating neurons, and to a lesser extent reactive glial cells. It is a major component of the motile "growth cones" that form the tips of elongating axons. GAP43 also plays a role in axonal and dendritic filopodia induction.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Axonal membrane protein GAP-43; brain abundant, membrane attached signal protein 2; Calmodulin-binding protein P-57; growth accentuating protein 43; Growth-associated protein 43; MGC156666; Neuromodulin; neuron protein F1; Protein F1
Gene Aliases: B-50; Basp2; GAP-43; Gap43
UniProt ID: (Rat) P07936, (Mouse) P06837
Entrez Gene ID: (Rat) 29423, (Mouse) 14432
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support