Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 1
GEF-H1 activates Rho GTPases by promoting the exchange of GDP and GTP. Rho GTPases play a fundamental role in numerous cellular processes that are initiated by extracellular stimuli that work through G protein coupled receptors. GEF-H1 may be involved in epithelial barrier permeability, cell motility and polarization, dendritic spine morphology, antigen presentation, leukemic cell differentiation, cell cycle regulation, innate immune response, and cancer. Alternatively spliced transcript variants encoding different isoforms have been identified.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DKFZp547L106; DKFZp547P1516; FLJ45440; GEF-H1; Guanine nucleotide exchange factor H1; LBC'S first cousin; Lymphoid blast crisis-like 1; Microtubule-regulated Rho-GEF; Oncogene LFC; OTTHUMP00000018855; Proliferating cell nucleolar antigen p40; Rho guanine nucleotide exchange factor 2; Rho/Rac guanine nucleotide exchange factor (GEF) 2; Rhobin
Gene Aliases: AA408978; ARHGEF2; GEF; GEF-H1; GEFH1; KIAA0651; Lbcl1; Lfc; LFP40; mKIAA0651; P40
UniProt ID: (Human) D3DVA6, (Mouse) Q60875, (Rat) Q5FVC2
Entrez Gene ID: (Human) 9181, (Mouse) 16800, (Rat) 310635
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support