Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
Antibody detects endogenous levels of HCLS1 only when phosphorylated at Tyr397.
Substrate of the antigen receptor-coupled tyrosine kinase. Plays a role in antigen receptor signaling for both clonal expansion and deletion in lymphoid cells. May also be involved in the regulation of gene expression. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: Hematopoietic cell-specific LYN substrate 1; Hematopoietic lineage cell-specific protein; LckBP1; p75
Gene Aliases: AW213261; CTTNL; HCLS1; HS1; lckBP1; p75
UniProt ID: (Human) P14317, (Mouse) P49710
Entrez Gene ID: (Human) 3059, (Mouse) 15163, (Rat) 288077
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support