Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • HIPK1/HIPK2/HIPK3 Antibodies

          Invitrogen

          Phospho-HIPK1/HIPK2/HIPK3 (Tyr352, Tyr361, Tyr359) Polyclonal Antibody

          View all (2) HIPK1/HIPK2/HIPK3 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-HIPK1/HIPK2/HIPK3 (Tyr352, Tyr361, Tyr359) Polyclonal Antibody

          Product Details

          PA5-64584

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthesized peptide derived from human HIPK1/2/3 (Phospho-Tyr352/361/359)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          1 mg/mL

          Purification

          Affinity chromatography

          Storage buffer

          PBS, pH 7.4, with 50% glycerol

          Contains

          0.02% sodium azide

          Storage conditions

          -20°C

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2663930

          Product Specific Information

          Phospho-HIPK1/HIPK2/HIPK3 (Tyr352, Tyr361, Tyr359) Polyclonal Antibody detects endogenous levels of HIPK1/HIPK2/HIPK3 only when phosphorylated at Tyr352, Tyr361, Tyr359.

          Target Information

          The protein encoded by this gene belongs to the Ser/Thr family of protein kinases and HIPK subfamily. It phosphorylates homeodomain transcription factors and may also function as a co-repressor for homeodomain transcription factors. Alternative splicing results in four transcript variants encoding four distinct isoforms.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Androgen receptor-interacting nuclear protein kinase; ANPK; Fas-interacting serine/threonine-protein kinase; FIST; HCMV-interacting protein; hHIPk2; hipk1 {ECO:0000312|EMBL:ABO47653.1}; homeodomain interacting protein kinase 1-like protein; Homeodomain-interacting protein kinase 1; Homeodomain-interacting protein kinase 2; Homeodomain-interacting protein kinase 3; Homolog of protein kinase YAK1; Nbak3; nuclear body associated kinase 2b; Nuclear body-associated kinase 1; Nuclear body-associated kinase 2; Nuclear body-associated kinase 3; nuclear serine/threonine protein kinase; Protein kinase Myak; Sialophorin tail-associated nuclear serine/threonine-protein kinase

          View more View less

          Gene Aliases: 1110014O20Rik; 1110062K04Rik; ANPK; B230339E18Rik; DYRK6; Fist; FIST3; HIPK1; HIPK2; HIPK3; KIAA0630; mir-1902; MYAK; Nbak1; NBAK2; PKY; PRO0593; Stank; YAK1

          View more View less

          UniProt ID: (Human) Q86Z02, (Human) Q9H2X6, (Human) Q9H422, (Mouse) O88904, (Mouse) Q9QZR5, (Mouse) Q9ERH7, (Rat) O88850

          View more View less

          Entrez Gene ID: (Human) 204851, (Human) 28996, (Human) 10114, (Rat) 365895, (Mouse) 15257, (Rat) 362342, (Mouse) 15258, (Mouse) 15259, (Rat) 83617

          View more View less

          Function(s)
          protein serine/threonine kinase activity protein binding ATP binding RNA polymerase II activating transcription factor binding RNA polymerase II transcription coactivator activity transcription corepressor activity protein kinase activity SMAD binding virion binding nucleotide binding DNA binding kinase activity transferase activity transferase activity, transferring phosphorus-containing groups
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter transcription, DNA-templated regulation of transcription, DNA-templated protein phosphorylation apoptotic process mRNA transcription phosphorylation peptidyl-serine phosphorylation peptidyl-threonine phosphorylation negative regulation of apoptotic process positive regulation of DNA binding negative regulation of JUN kinase activity smoothened signaling pathway positive regulation of cell proliferation anterior/posterior pattern specification regulation of tumor necrosis factor-mediated signaling pathway retina layer formation neuron differentiation intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator embryonic camera-type eye morphogenesis embryonic retina morphogenesis in camera-type eye lens induction in camera-type eye iris morphogenesis endothelial cell apoptotic process extrinsic apoptotic signaling pathway positive regulation of protein phosphorylation DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator transforming growth factor beta receptor signaling pathway adult walking behavior adult locomotory behavior positive regulation of transforming growth factor beta receptor signaling pathway negative regulation of BMP signaling pathway positive regulation of protein binding negative regulation of neuron apoptotic process positive regulation of angiogenesis positive regulation of transcription, DNA-templated positive regulation of transcription from RNA polymerase II promoter positive regulation of JNK cascade voluntary musculoskeletal movement positive regulation of sequence-specific DNA binding transcription factor activity SMAD protein signal transduction eye development modulation by virus of host morphology or physiology erythrocyte differentiation PML body organization regulation of cell cycle cellular response to hypoxia intrinsic apoptotic signaling pathway regulation of signal transduction by p53 class mediator adherens junction assembly definitive hemopoiesis cellular response to DNA damage stimulus negative regulation of nucleic acid-templated transcription
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-74594944d-pk6zc:80/100.66.130.62:80.
          git-commit: ab2b2261c2971e97b1ae408e3ad10520b8413932
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0