Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PDGFRB (CD140b) Antibodies

          Invitrogen

          Phospho-PDGFRB (Tyr751) Polyclonal Antibody

          View all (93) PDGFRB (CD140b) antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-PDGFRB (Tyr751) Polyclonal Antibody

          • Antibody Testing Data (3)
          Phospho-PDGFRB (Tyr751) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-PDGFRB (Tyr751) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 3

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-PDGFRB (Tyr751) Antibody (PA5-85805) in ICC/IF

          Immunocytochemistry analysis of Phospho-PDGFRB (Tyr751) in Mock and treated NIH/3T3 cells fixed in 4% paraformaldehyde at RT for 15 min. Samples were incubated with Phospho-PDGFRB (Tyr751) polyclonal antibody (Product # PA5-85805) using a dilution of 1:1,000 (green). Blue: Hoechst 33342 staining. Scale bar = 10 μm. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-PDGFRB (Tyr751) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-PDGFRB (Tyr751) Antibody in Western Blot (WB)
          Phospho-PDGFRB (Tyr751) Antibody in Western Blot (WB)

          Product Details

          PA5-85805

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:3,000
          -

          Immunocytochemistry (ICC/IF)

          1:100-1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Carrier-protein conjugated synthetic peptide surrounding phospho Tyr751 of human PDGF Receptor beta. The exact sequence is proprietary.
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.12 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS, pH 7, with 20% glycerol, 1% BSA

          Contains

          0.025% ProClin 300

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2792941

          Product Specific Information

          Keep as concentrated solution.

          Predicted reactivity: Bovine (100%).

          Positive Control: PDGF treated NIH-3T3.

          Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

          Target Information

          PDGFRB is a receptor tyrosine kinase and has been implicated in atherogenesis and in cell transformation by the SIS oncogene. These growth factors are mitogens for cells of mesenchymal origin. The identity of the growth factor bound to a receptor monomer determines whether the functional receptor is a homodimer or a heterodimer, composed of both platelet-derived growth factor receptor alpha and beta polypeptides. The gene is flanked on chromosome 5 by the genes for granulocyte-macrophage colony-stimulating factor and macrophage-colony stimulating factor receptor; all three genes may be implicated in the 5-q syndrome. A translocation between chromosomes 5 and 12, that fuses PDGFRB (CD140b) to that of the translocation, ETV6, leukemia gene, results in chronic myeloproliferative disorder with eosinophilia.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Beta platelet-derived growth factor receptor; Beta-type platelet-derived growth factor receptor; CD140 antigen-like family member B; CD140b; CD140b antigen; PDGF beta chain; PDGF Receptor beta; PDGF-R-beta; PDGFR-1; PDGFR-beta; Platelet-derived growth factor receptor 1; Platelet-derived growth factor receptor beta; platelet-derived growth factor receptor beta variant 1; platelet-derived growth factor receptor, beta polypeptide

          View more View less

          Gene Aliases: AI528809; CD140B; IBGC4; IMF1; JTK12; KOGS; PDGFR; PDGFR-1; PDGFR1; PDGFRB; PENTT

          View more View less

          UniProt ID: (Human) P09619, (Mouse) P05622

          View more View less

          Entrez Gene ID: (Human) 5159, (Mouse) 18596

          View more View less

          Function(s)
          protein tyrosine kinase activity platelet activating factor receptor activity platelet-derived growth factor-activated receptor activity platelet-derived growth factor beta-receptor activity Ras guanyl-nucleotide exchange factor activity receptor binding platelet-derived growth factor receptor binding protein binding ATP binding protein kinase binding vascular endothelial growth factor binding phosphatidylinositol 3-kinase binding phosphatidylinositol-4,5-bisphosphate 3-kinase activity platelet-derived growth factor binding nucleotide binding protein kinase activity transmembrane receptor protein tyrosine kinase activity signal transducer activity kinase activity transferase activity transferase activity, transferring phosphorus-containing groups
          Process(es)
          in utero embryonic development kidney development tissue homeostasis glycosaminoglycan biosynthetic process protein phosphorylation nitrogen compound metabolic process chemotaxis signal transduction transmembrane receptor protein tyrosine kinase signaling pathway multicellular organism development positive regulation of cell proliferation positive regulation of phospholipase C activity positive regulation of phosphatidylinositol 3-kinase signaling positive regulation of smooth muscle cell migration phosphorylation cell migration peptidyl-tyrosine phosphorylation hemopoiesis adrenal gland development positive regulation of cell migration positive regulation of phosphoprotein phosphatase activity regulation of actin cytoskeleton organization positive regulation of collagen biosynthetic process cell migration involved in vasculogenesis intracellular signal transduction metanephric mesenchymal cell migration platelet-derived growth factor receptor-beta signaling pathway positive regulation of metanephric mesenchymal cell migration by platelet-derived growth factor receptor-beta signaling pathway aorta morphogenesis cellular response to platelet-derived growth factor stimulus positive regulation of cell proliferation by VEGF-activated platelet derived growth factor receptor signaling pathway negative regulation of apoptotic process positive regulation of MAP kinase activity positive regulation of phosphatidylinositol 3-kinase activity positive regulation of mitotic nuclear division phosphatidylinositol metabolic process protein autophosphorylation platelet-derived growth factor receptor signaling pathway phosphatidylinositol-mediated signaling positive regulation of smooth muscle cell proliferation skeletal system morphogenesis smooth muscle tissue development regulation of peptidyl-tyrosine phosphorylation positive regulation of chemotaxis cardiac myofibril assembly cell chemotaxis cardiac vascular smooth muscle cell differentiation cell migration involved in coronary angiogenesis retina vasculature development in camera-type eye positive regulation of ERK1 and ERK2 cascade smooth muscle cell chemotaxis metanephric glomerular mesangial cell proliferation involved in metanephros development metanephric glomerular capillary formation positive regulation of calcium ion import positive regulation of reactive oxygen species metabolic process positive regulation of DNA biosynthetic process MAPK cascade G-protein coupled receptor signaling pathway response to toxic substance regulation of phosphatidylinositol 3-kinase signaling response to estradiol response to retinoic acid response to fluid shear stress wound healing response to hydrogen peroxide positive regulation of GTPase activity response to estrogen phosphatidylinositol phosphorylation inner ear development response to hyperoxia metanephric mesenchyme development metanephric comma-shaped body morphogenesis metanephric S-shaped body morphogenesis
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-2w6sl:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0