Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PI3K p85/p55 Antibodies

          Invitrogen

          Phospho-PI3K p85/p55 (Tyr467, Tyr199, Tyr464) Polyclonal Antibody

          View all (13) PI3K p85/p55 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-PI3K p85/p55 (Tyr467, Tyr199, Tyr464) Polyclonal Antibody

          • Antibody Testing Data (1)
          Phospho-PI3K p85/p55 (Tyr467, Tyr199, Tyr464) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-PI3K p85/p55 (Tyr467, Tyr199, Tyr464) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-PI3K p85/p55 (Tyr467, Tyr199, Tyr464) Antibody (PA5-121306) in WB

          Western blot analysis of Phospho-PI3K p85/p55 (Tyr467, Tyr199, Tyr464) in extracts of HeLa cells. Samples were incubated with Phospho-PI3K p85/p55 (Tyr467, Tyr199, Tyr464) Polyclonal antibody (Product # PA5-121306) using a dilution of 1:1,000, followed by HRP Goat Anti-Rabbit IgG (H+L) secondary antibody at a dilution of 1:10,000. Hela cells were treated by CIP (20 µL/400 µL) at 37°C for 1 hour. Lysates/pro... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-PI3K p85/p55 (Tyr467, Tyr199, Tyr464) Antibody in Western Blot (WB)

          Product Details

          PA5-121306

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:500-1:2,000
          -

          ELISA (ELISA)

          1 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic phosphorylated peptide around Y467/Y199/Y464 of human p85-alpha (NP_8526641)
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.62 mg/mL

          Purification

          Affinity Chromatography

          Storage buffer

          PBS, pH 7.3, with 50% glycerol

          Contains

          0.05% ProClin 300

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2914878

          Product Specific Information

          Positive test controls include: HeLa, C2C12.

          Immunogen sequence: DRLYEE

          Target Information

          Phosphoinositide 3-kinase (PI3K, phospho PI3K) Binds to activated (phosphorylated) protein-Tyr kinases, through its SH2 domain, and acts as an adapter, mediating the association of the p110 catalytic unit to the plasma membrane. Phospho PI3K is necessary for the insulin-stimulated increase in glucose uptake and glycogen synthesis in insulin-sensitive tissues. During insulin stimulation, it also binds to IRS-1. Growth factors and hormones trigger this phosphorylation event, which in turn coordinates cell growth, cell cycle entry, cell migration, and cell survival.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: OTTHUMP00000009783; OTTHUMP00000009784; p55PIK; phosphatidylinositol 3 kinase, regulatory subunit, polypeptide 3 (p55); Phosphatidylinositol 3-kinase 55 kDa regulatory subunit gamma; Phosphatidylinositol 3-kinase 85 kDa regulatory subunit alpha; Phosphatidylinositol 3-kinase 85 kDa regulatory subunit beta; phosphatidylinositol 3-kinase p55 subunit; phosphatidylinositol 3-kinase regulartory subunit polypeptide 2; Phosphatidylinositol 3-kinase regulatory subunit alpha; Phosphatidylinositol 3-kinase regulatory subunit beta; Phosphatidylinositol 3-kinase regulatory subunit gamma; phosphatidylinositol 3-kinase, regulartory subunit, polypeptide 2; phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha); phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 2 (p85 beta); phosphatidylinositol 3-kinase, regulatory subunit, polypeptide 3 (p55, gamma); phosphatidylinositol 3-kinase-associated p-85 alpha; Phosphoinositide 3-kinase p85 (other splicing variants: p55 and p50); Phosphoinositide 3-kinase, regulatory subunit, polypeptide 1 (p85 alpha); phosphoinositide-3-kinase regulatory subunit; phosphoinositide-3-kinase regulatory subunit alpha; phosphoinositide-3-kinase regulatory subunit beta; phosphoinositide-3-kinase, regulatory subunit 1 (alpha); phosphoinositide-3-kinase, regulatory subunit 2 (beta); phosphoinositide-3-kinase, regulatory subunit 3 (gamma); PI3-kinase p85 subunit alpha; PI3-kinase p85 subunit beta; PI3-kinase p85 subunit gamma; PI3-kinase regulatory subunit alpha; PI3-kinase regulatory subunit beta; PI3-kinase regulatory subunit gamma; PI3-kinase subunit p55-gamma; PI3-kinase subunit p85-alpha; PI3-kinase subunit p85-beta; PI3K regulatory subunit alpha; PI3K regulatory subunit beta; PI3K regulatory subunit gamma; polypeptide 3; ptdIns-3-kinase p85-alpha; ptdIns-3-kinase p85-beta; ptdIns-3-kinase p85-gamma; ptdIns-3-kinase regulatory subunit alpha; ptdIns-3-kinase regulatory subunit beta; ptdIns-3-kinase regulatory subunit gamma; ptdIns-3-kinase regulatory subunit p55-gamma; PtdIns-3-kinase regulatory subunit p85-al; ptdIns-3-kinase regulatory subunit p85-alpha; ptdIns-3-kinase regulatory subunit p85-beta; RP4-533D7.2

          View more View less

          Gene Aliases: AA414954; AGM7; GRB1; IMD36; MPPH; MPPH1; p50alpha; p55; p55-GAMMA; p55alpha; p55PIK; p85; p85-ALPHA; p85-BETA; p85alpha; P85B; p85beta; PI3K; PI3KA; PIK3R1; PIK3R2; PIK3R3

          View more View less

          UniProt ID: (Human) P27986, (Human) O00459, (Human) Q92569, (Mouse) P26450, (Rat) Q63787, (Mouse) O08908, (Rat) Q63789, (Mouse) Q64143

          View more View less

          Entrez Gene ID: (Human) 5295, (Human) 5296, (Human) 8503, (Mouse) 18708, (Rat) 25513, (Mouse) 18709, (Rat) 29741, (Rat) 60664, (Mouse) 18710

          View more View less

          Function(s)
          transmembrane receptor protein tyrosine kinase adaptor activity insulin receptor binding insulin-like growth factor receptor binding neurotrophin TRKA receptor binding protein binding transcription factor binding 1-phosphatidylinositol-3-kinase activity protein phosphatase binding phosphatidylinositol 3-kinase regulator activity phosphatidylinositol 3-kinase regulatory subunit binding ErbB-3 class receptor binding phosphatidylinositol 3-kinase binding insulin binding insulin receptor substrate binding phosphatidylinositol-4,5-bisphosphate 3-kinase activity 1-phosphatidylinositol-3-kinase regulator activity protein heterodimerization activity GTPase activator activity receptor tyrosine kinase binding receptor binding platelet-derived growth factor receptor binding calmodulin binding protein C-terminus binding protein kinase binding protein domain specific binding estrogen receptor binding ubiquitin protein ligase binding ATPase binding phosphoprotein binding kinase activity
          Process(es)
          insulin receptor signaling pathway phosphorylation regulation of phosphatidylinositol 3-kinase activity phosphatidylinositol biosynthetic process phosphatidylinositol-3-phosphate biosynthetic process positive regulation of glucose import in response to insulin stimulus cellular glucose homeostasis regulation of autophagy phosphatidylinositol 3-kinase signaling regulation of phosphatidylinositol 3-kinase signaling protein transport cellular response to insulin stimulus response to endoplasmic reticulum stress Fc-epsilon receptor signaling pathway Fc-gamma receptor signaling pathway involved in phagocytosis positive regulation of transcription factor import into nucleus positive regulation of GTPase activity positive regulation of transcription from RNA polymerase II promoter phosphatidylinositol phosphorylation vascular endothelial growth factor receptor signaling pathway phosphatidylinositol-mediated signaling T cell receptor signaling pathway leukocyte migration regulation of small GTPase mediated signal transduction positive regulation of protein phosphorylation negative regulation of cell-matrix adhesion glucose metabolic process protein phosphorylation transport negative regulation of cell adhesion signal transduction extrinsic apoptotic signaling pathway via death domain receptors intrinsic apoptotic signaling pathway in response to DNA damage negative regulation of heart rate positive regulation of gene expression negative regulation of muscle cell apoptotic process negative regulation of cell-cell adhesion B cell differentiation positive regulation of cell migration positive regulation of tumor necrosis factor production response to insulin positive regulation of RNA splicing cellular response to UV negative regulation of apoptotic process positive regulation of myoblast differentiation negative regulation of osteoclast differentiation negative regulation of blood pressure negative regulation of proteolysis insulin-like growth factor receptor signaling pathway negative regulation of smooth muscle cell proliferation protein stabilization response to glucocorticoid regulation of stress fiber assembly NFAT protein import into nucleus response to cAMP growth hormone receptor signaling pathway regulation of establishment of protein localization to plasma membrane positive regulation of endoplasmic reticulum unfolded protein response epidermal growth factor receptor signaling pathway viral process platelet activation T cell costimulation ERBB2 signaling pathway positive regulation of glucose import positive regulation of establishment of protein localization to plasma membrane regulation of actin filament polymerization regulation of phosphorylation positive regulation of cell adhesion response to yeast aging response to nutrient response to fructose response to iron(II) ion response to estradiol response to progesterone response to testosterone response to drug response to amino acid response to ethanol response to fatty acid response to growth factor cellular response to fatty acid response to dexamethasone
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0