Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • PLCG1 Antibodies

          Invitrogen

          Phospho-PLCG1 (Tyr775) Polyclonal Antibody

          View all (41) PLCG1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-PLCG1 (Tyr775) Polyclonal Antibody

          • Antibody Testing Data (1)
          Phospho-PLCG1 (Tyr775) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-PLCG1 (Tyr775) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-PLCG1 (Tyr775) Antibody (PA5-143744) in WB

          Western blot of Phospho-PLCG1 (Tyr775) immunoprecipitation from human jurkat cells untreated (lanes 1 & 3) or treated with pervanadate (1 mM) for 30 minutes (lanes 2,4,5,6). Immunoprecipitation was performed with PLCG1 monoclonal antibody (Product # MA5-47678). The blots were probed with anti-PLCgamma1 (lanes 1 & 2) and Phospho-PLCG1 (Tyr775) polyclonal antibody (Product # PA5-143744) (lanes 3-6). The latte... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-PLCG1 (Tyr775) Antibody in Western Blot (WB)

          Product Details

          PA5-143744

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:100
          -

          Immunoprecipitation (IP)

          1:100
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          PLCgamma1 (Tyr-775) synthetic peptide (coupled to carrier protein) corresponding to amino acids around tyrosine 775 in human PLCgamma1. This sequence has high homology to the conserved site in rat and mouse PLCgamma1, and has low homology to PLCgamma2.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA, 50% glycerol

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2942972

          Product Specific Information

          This antibody was cross-adsorbed to a non-specific phospho-tyrosine peptide then affinity-purified using phospho-PLCgamma1 (Tyr-775) peptide. The antibody detects a 150 kDa* protein in human Jurkat and A431 cells treated with pervanadate, but is not observed in untreated cells.

          Target Information

          The mammalian Phospholipase Cg (PLCg) family has two closely related proteins, PLCg1 and PLCg2. The PLCg isozymes have the core structure domains and a unique array of domains containing an additional PH domain, two SH2 domains and one SH3 domain. In response to extracellular stimuli, such as hormones and growth factors, receptor tyrosine kinases (RTKs) phosphorylate and activate PLCg. Activated PLCg catalyzes hydrolysis of phosphatidylinositol 4,5-bisphosphate (PIP2) to produce the metabolic second messenger molecules inositol 1,4,5-triphosphate (IP3) and diacylglycerol (DAG).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: 1-phosphatidyl-D-myo-inositol-4,5-bisphosphate; 1-phosphatidylinositol 4,5-bisphosphate phosphodiesterase gamma-1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma 1; 1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-1; cell differentiation and embryonic development; EC 3.1.4.11; inositoltrisphosphohydrolase; monophosphatidylinositol phosphodiesterase; OTTHUMP00000178982; phosphatidylinositol phospholipase C; phosphoinositidase C; phosphoinositide phospholipase C; Phosphoinositide phospholipase C-gamma-1; phospholipase C, gamma 1 (formerly subtype 148); phospholipase C-1; phospholipase C-148; Phospholipase C-gamma-1; Phospholipase C-II; Phospholipase C2; PLC gamma1; PLC-148; PLC-gamma-1; PLC-II; PLC2I; triphosphoinositide phosphodiesterase

          View more View less

          Gene Aliases: AI894140; Cded; NCKAP3; Plc-1; Plc-gamma1; PLC-II; PLC1; PLC148; Plcg-1; PLCG1; PLCgamma1; PPLCA

          View more View less

          UniProt ID: (Human) P19174, (Mouse) Q62077, (Rat) P10686

          View more View less

          Entrez Gene ID: (Human) 5335, (Mouse) 18803, (Rat) 25738

          View more View less

          Function(s)
          phosphatidylinositol phospholipase C activity phospholipase C activity receptor signaling protein activity neurotrophin TRKA receptor binding calcium ion binding protein binding protein kinase binding receptor tyrosine kinase binding glutamate receptor binding signal transducer activity insulin receptor binding phosphoric diester hydrolase activity hydrolase activity metal ion binding phosphoprotein binding
          Process(es)
          in utero embryonic development lipid metabolic process calcium ion transport signal transduction epidermal growth factor receptor signaling pathway protein secretion phospholipid catabolic process positive regulation of epithelial cell migration lipid catabolic process cell migration calcium-mediated signaling inositol trisphosphate biosynthetic process intracellular signal transduction positive regulation of blood vessel endothelial cell migration positive regulation of angiogenesis T cell receptor signaling pathway positive regulation of release of sequestered calcium ion into cytosol cellular response to epidermal growth factor stimulus regulation of store-operated calcium channel activity response to gravity response to organonitrogen compound response to hydrogen peroxide response to morphine activation of MAPKK activity viral process Fc-epsilon receptor signaling pathway Fc-gamma receptor signaling pathway involved in phagocytosis inositol phosphate metabolic process leukocyte migration
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0