Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • Profilin 1 Antibodies

          Invitrogen

          Phospho-Profilin 1 (Ser138) Polyclonal Antibody

          View all (26) Profilin 1 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-Profilin 1 (Ser138) Polyclonal Antibody

          • Antibody Testing Data (2)
          Phospho-Profilin 1 (Ser138) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-Profilin 1 (Ser138) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-Profilin 1 (Ser138) Antibody (PA5-143752) in WB

          Western blot of Phospho-Profilin 1 (Ser138) in Jurkat stimulated with calyculin A (100 nM) for 30 min (lanes 1-6). The blots were probed with anti-Profilin (a.a. 132-140) (lanes 1-3) or Phospho-Profilin 1 (Ser138) polyclonal antibody (Product # PA5-143752) (lanes 4-6). Both antibodies were used in the absence (lanes 1 & 4) or presence of unphosphorylated Profilin (Ser-138) (lanes 2 & 5) or phospho-Profilin ... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-Profilin 1 (Ser138) Antibody in Western Blot (WB)
          Phospho-Profilin 1 (Ser138) Antibody in Western Blot (WB)

          Product Details

          PA5-143752

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Phospho-Profilin (Ser138) synthetic peptide (coupled to carrier protein) that includes amino acids surrounding Ser138 in human Profilin-1. This sequence is conserved in rat and mouse Profilin-1. The serine site has some homology to the conserved site (Ser138) in Profilin-2a and Profilin-2b, but the site is not conserved in Profiin-3 and Profilin-4.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          ~0.5 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA, 50% glycerol

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2942980

          Product Specific Information

          This antibody was cross-adsorbed to unphosphorylated Profilin (Ser-138) then affinity purified using Profilin (Ser-138) peptide (without carrier). The antibody detects a 15 kDa band corresponding to the apparent molecular mass of Profilin-1 in human Jurkat and A431 cells, as well as rat PC12 cells. This reactivity is decreased by lambda phosphatase treatment, and is blocked by phospho-profilin (Ser-138) blocking peptide.

          Target Information

          Profilin is an ubiquitous small (12-15 kDa) phosphoinositides and poly-L-proline binding protein that plays a role in both signal transduction pathways and actin filament dynamics. There are two mammalian profilins with similar biochemical properties but different expression patterns. Profilin I appears to be highly expressed in most tissues except for skeletal muscle. Profilin II is predominantly expressed in brain and at lower levels in skeletal muscle, uterus and kidney. Profilin is a mainly cytosolic protein with higher concentrations in dynamic membrane areas like the leading edge and ruffling membranes.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: actin binding protein; Epididymis tissue protein Li 184a; Profilin I; Profilin-1

          View more View less

          Gene Aliases: ALS18; Pfn; PFN1

          View more View less

          UniProt ID: (Human) P07737, (Rat) P62963, (Mouse) P62962

          View more View less

          Entrez Gene ID: (Human) 5216, (Rat) 64303, (Mouse) 18643

          View more View less

          Function(s)
          adenyl-nucleotide exchange factor activity actin binding actin monomer binding receptor binding protein binding phosphatidylinositol-4,5-bisphosphate binding Rho GTPase binding poly(A) RNA binding proline-rich region binding cadherin binding involved in cell-cell adhesion
          Process(es)
          neural tube closure regulation of transcription from RNA polymerase II promoter actin polymerization or depolymerization response to organic substance positive regulation of epithelial cell migration negative regulation of actin filament polymerization positive regulation of actin filament polymerization negative regulation of actin filament bundle assembly positive regulation of ATPase activity positive regulation of transcription from RNA polymerase II promoter positive regulation of viral transcription positive regulation of DNA metabolic process positive regulation of stress fiber assembly negative regulation of stress fiber assembly cellular response to growth factor stimulus positive regulation of ruffle assembly actin cytoskeleton organization positive regulation of actin filament bundle assembly protein stabilization Wnt signaling pathway, planar cell polarity pathway cell-cell adhesion regulation of actin polymerization or depolymerization sequestering of actin monomers
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0