Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Phospho-RARA/RARB (Ser96) Polyclonal Antibody detects endogenous levels of RARA/RARB only when phosphorylated at Ser96.
Retinoic acid, a metabolite of vitamin A, is necessary for normal organogenesis but acts as a teratogen at high levels during embryonic and fetal development. Retinoic acid functions through its interaction with the nuclear protein, retinoic acid receptor (RAR). RAR belongs to the steroid and thyroid hormone superfamily of nuclear receptor proteins which exert their effects by binding to specific DNA response elements, thus regulating gene expression in target cells. RAR exists as three major subtypes: alpha, beta and gamma.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: HBV-activated protein; hepatitis B virus activated protein; Nuclear receptor subfamily 1 group B member 1; Nuclear receptor subfamily 1 group B member 2; nucleophosmin-retinoic acid receptor alpha fusion protein NPM-RAR long form; RAR alpha 1; RAR-alpha; RAR-beta; RAR-epsilon; retinoic acid; retinoic acid nuclear receptor alpha variant 1; retinoic acid nuclear receptor alpha variant 2; retinoic acid receptor; Retinoic acid receptor alpha; Retinoic acid receptor beta; retinoic acid receptor, alpha polypeptide; retinoic acid receptor, beta polypeptide
Gene Aliases: A830025K23; HAP; MCOPS12; NR1B1; NR1B2; RAR; RARA; RARalpha1; RARB; RARbeta1; RRB2
UniProt ID: (Human) P10276, (Human) P10826, (Mouse) P11416, (Mouse) P22605
Entrez Gene ID: (Human) 5914, (Human) 5915, (Rat) 24705, (Mouse) 19401, (Rat) 24706, (Mouse) 218772
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support