Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • c-Rel Antibodies

          AbboMax

          Phospho-Rel (Ser503) Polyclonal Antibody

          View all (24) c-Rel antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-Rel (Ser503) Polyclonal Antibody

          • Antibody Testing Data (1)
          Phospho-Rel (Ser503) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-Rel (Ser503) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-Rel (Ser503) Antibody (500-12024) in WB

          Western Blot: The cell lysate derived from PMA treated Jurkat cells was immunoprobed by Rabbit anti-c-Rel (pS503) (Cat#500-12024) at 1:500. A major immunoreactive band is observed around ~68 kDa. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-Rel (Ser503) Antibody in Western Blot (WB)

          Product Details

          500-12024

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          0.1-1 µg/mL
          -

          Immunohistochemistry (Paraffin) (IHC (P))

          2-10 µg/mL
          -

          Immunoprecipitation (IP)

          2-5 µg/mL
          -
          Product Specifications

          Species Reactivity

          Human, Mouse

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide corresponding to the epitope -TTSSDS-at a phosphorylation site Ser503 of human c-Rel protein
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.5 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          25mM tris with proprietary stabilizer

          Contains

          0.01% sodium azide

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.

          Shipping conditions

          Wet ice

          Product Specific Information

          Positive control: PMA-treated Jurkat cells

          Cellular location: Nucleus, Cytoplasm.

          Target Information

          The REL gene encodes c-Rel, a transcription factor that is a member of the Rel/NFKB family, which also includes RELA., RELB (604758), NFKB1, and NFKB2. These proteins are related through a highly conserved N-terminal region termed the 'Rel domain, ' which is responsible for DNA binding, dimerization, nuclear localization, and binding to the NFKB inhibitor (Belguise and Sonenshein, 2007).

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: BCD541; C-Rel protein; C-Rel proto-oncogene protein; gemin 1; gemin-1; oncogene REL, avian reticuloendotheliosis; Proto-oncogene c-Rel; SMA; SMA1; SMA2; SMA3; SMA4; SMN; SMN2; SMNT; v-rel avian reticuloendotheliosis viral oncogene homolog

          View more View less

          Gene Aliases: C-Rel; REL

          View more View less

          UniProt ID: (Human) Q04864

          View more View less

          Entrez Gene ID: (Human) 5966, (Mouse) 19696

          View more View less

          Function(s)
          RNA polymerase II distal enhancer sequence-specific DNA binding transcriptional activator activity, RNA polymerase II distal enhancer sequence-specific binding chromatin binding transcription factor activity, sequence-specific DNA binding protein binding RNA polymerase II regulatory region sequence-specific DNA binding RNA polymerase II transcription factor activity, sequence-specific DNA binding DNA binding sequence-specific DNA binding
          Process(es)
          negative regulation of transcription from RNA polymerase II promoter transcription from RNA polymerase II promoter inflammatory response I-kappaB kinase/NF-kappaB signaling negative regulation of gene expression negative regulation of interferon-beta production response to cytokine NIK/NF-kappaB signaling positive regulation of I-kappaB kinase/NF-kappaB signaling innate immune response positive regulation of transcription from RNA polymerase II promoter negative regulation of neuron death cytokine production transcription, DNA-templated regulation of transcription, DNA-templated positive regulation of interleukin-12 biosynthetic process positive regulation of transcription, DNA-templated
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-67c77f6dc7-n6nf2:80/100.66.135.186:80.
          git-commit: 545e792480003146e539d624bd2a21b51d198228
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0