Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
FIGURE: 1 / 2
The antibody detects a 150 kDa* full length recombinant human SSH1L phosphoprotein and this reactivity is eliminated with lambda phosphatase treatment. The antibody also detects SSH1L on immunoblots of rat PC12 and mouse C2C12 cells that were stimulated with calyculin A.
This gene encodes a member of the SWI/SNF family, whose members have helicase and ATPase activities and are thought to regulate transcription of certain genes by altering the chromatin structure around those genes. The encoded protein is part of the large ATP-dependent chromatin remodeling complex SNF/SWI, which is required for transcriptional activation of genes normally repressed by chromatin. It possesses at least two conserved domains that could be important for its function. First, it has a DNA-binding domain that can specifically bind an AT-rich DNA sequence known to be recognized by a SNF/SWI complex at the beta-globin locus. Second, the C-terminus of the protein can stimulate glucocorticoid receptor-dependent transcriptional activation. It is thought that the protein encoded by this gene confers specificity to the SNF/SWI complex and may recruit the complex to its targets through either protein-DNA or protein-protein interactions. Two transcript variants encoding different isoforms have been found for this gene.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: hSSH-1L; Protein phosphatase Slingshot homolog 1; slingshot 1; slingshot 1L; slingshot homolog 1; slingshot-like 1; SSH-1L; SSH-like protein 1
Gene Aliases: AW551225; Gm1394; Gm1395; KIAA1298; mSSH-1L; SSH-1; SSH1; SSH1L
UniProt ID: (Human) Q8WYL5, (Mouse) Q76I79
Entrez Gene ID: (Human) 54434, (Mouse) 231637, (Rat) 304580
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support