Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
FIGURE: 1 / 6
Antibody detects endogenous levels of TOPBP1 only when phosphorylated at Ser1159.
This gene encodes a binding protein which interacts with the C-terminal region of topoisomerase II beta. This interaction suggests a supportive role for this protein in the catalytic reactions of topoisomerase II beta through transient breakages of DNA strands.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: DNA topoisomerase 2-binding protein 1; DNA topoisomerase II-beta-binding protein 1; DNA topoisomerase II-binding protein 1; TOPBP 1; TopBP1; topoisomerase (DNA) II beta binding protein; topoisomerase (DNA) II binding protein 1
Gene Aliases: 1110031N14Rik; 2810429C13Rik; AI256758; D430026L04Rik; KIAA0259; mKIAA0259; RGD1562949; TOP2BP1; TOPBP1
UniProt ID: (Human) Q92547, (Mouse) Q6ZQF0
Entrez Gene ID: (Human) 11073, (Mouse) 235559, (Rat) 315969
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support