Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • TRPM7 Antibodies

          Invitrogen

          Phospho-TRPM7 (Ser1513) Polyclonal Antibody

          View all (30) TRPM7 antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-TRPM7 (Ser1513) Polyclonal Antibody

          • Antibody Testing Data (1)
          Phospho-TRPM7 (Ser1513) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.
          Phospho-TRPM7 (Ser1513) Antibody in Western Blot (WB)
          Group 53 Created with Sketch.

          FIGURE: 1 / 1

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-TRPM7 (Ser1513) Antibody (PA5-143779) in WB

          Western blot of Phospho-TRPM7 (Ser1513) in human auto phosphorylated TRPM7 C-terminal kinase domain (lanes 1-6). The blot was treated with lambda phosphatase to dephosphorylate TRPM7 phosphosites (lanes 2, 4, & 6). The blot was probed with rabbit Phospho-TRPM7 (Ser1513) polyclonal antibody (Product # PA5-143779) (lanes 1 & 2), anti-TRPM7 (a.a.1484-1497) antibody (lanes 3 & 4), or anti-TRPM7 (Ser1493), (lane... View More {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-TRPM7 (Ser1513) Antibody in Western Blot (WB)

          Product Details

          PA5-143779

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          Assay-dependent
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Phospho-TRPM7 (Ser-1513) synthetic peptide (coupled to carrier) corresponding to amino acids surrounding Ser-1513 in human TRPM7. This site is well conserved in rat and mouse TRPM7, but has low homology to other TRPM family members.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.25 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA, 50% glycerol

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          RRID

          AB_2943007

          Product Specific Information

          The antibody was cross-adsorbed to unphosphorylated TRPM7 (Ser-1513) peptide then affinity purified using phospho-TRPM7 (Ser-1513) peptide. This antibody detects a 220 kDa* protein on SDS-PAGE immunoblots of human MDA-MB-231 and rat PC12 cells stimulated with calyculin A, as well as detects a 70 kDa autophosphorylated recombinant kinase domain of human TRPM7. These reactivities are not observed after lambda phosphatase treatment to dephosphorylate TRPM7.

          Target Information

          TRPCs, mammalian homologs of the Drosophila transient receptor potential (trp) protein, are ion channels that are thought to mediate capacitative calcium entry into the cell. TRP-PLIK is a protein that is both an ion channel and a kinase. As a channel, it conducts calcium and monovalent cations to depolarize cells and increase intracellular calcium. As a kinase, it is capable of phosphorylating itself and other substrates. The kinase activity is necessary for channel function, as shown by its dependence on intracellular ATP and by the kinase mutants.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: Channel kinase 1; Channel-kinase 1; FLJ20117; Long transient receptor potential channel 7; long transient receptor potential channel 7 {ECO:0000250|UniProtKB:Q96QT4}; LTRPC ion channel family member 7; LTrpC-7; LTrpC7 {ECO:0000250|UniProtKB:Q96QT4}; tr; Transient receptor potential cation channel subfamily M member 7; transient receptor potential cation channel subfamily M member 7 {ECO:0000312|RGD:620053}; transient receptor potential cation channel, subfamily M, member 7; transient receptor potential M7; Transient receptor potential-phospholipase C-interacting kinase; transient receptor potential-phospholipase C-interacting kinase {ECO:0000303|PubMed:11161216}; transient receptor potential-related protein, ChaK; TRMP7; TRP PLIK; TRP-PLIK; TRP-PLIK {ECO:0000303|PubMed:11161216}; trpm7 {ECO:0000312|RGD:620053}

          View more View less

          Gene Aliases: 2310022G15Rik; 4833414K03Rik; 5033407O22Rik; ALSPDC; CHAK; CHAK1; Ltpr7; LTrpC-7; LTRPC7; TRP-PLIK; TRPM7; TRPPLIK

          View more View less

          UniProt ID: (Human) Q96QT4, (Mouse) Q923J1, (Rat) Q925B3

          View more View less

          Entrez Gene ID: (Human) 54822, (Mouse) 58800, (Rat) 679906

          View more View less

          Function(s)
          actin binding protein serine/threonine kinase activity calcium channel activity ATP binding myosin binding metal ion binding nucleotide binding protein kinase activity ion channel activity cation channel activity protein binding kinase activity transferase activity transferase activity, transferring phosphorus-containing groups
          Process(es)
          cellular magnesium ion homeostasis calcium-dependent cell-matrix adhesion actomyosin structure organization protein autophosphorylation protein tetramerization necroptotic process calcium ion transmembrane transport protein phosphorylation transport ion transport cation transport calcium ion transport memory programmed cell death phosphorylation positive regulation of apoptotic process transmembrane transport response to radiation
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0