Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • TrkA/TrkB/TrkC Antibodies

          Osenses

          Phospho-TrkA/B/C (Tyr683, Tyr684) Polyclonal Antibody

          1 Reference
          View all (12) TrkA/TrkB/TrkC antibodies

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}

          {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoTitle}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-TrkA/B/C (Tyr683, Tyr684) Polyclonal Antibody

          Product Details

          OST00251W-100UL

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:300-1:2,000
          -

          Immunohistochemistry (IHC)

          1:300-1:2,000
          -

          Miscellaneous PubMed (MISC)

          -
          View 1 publication 1 publication
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / Ig

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          A synthetic peptide to phospho Y683 & Y684 of rat TrkA (and the corresponding phosphorylated sequences in TrkB and TrkC) conjugated to an immunogenic carrier protein was used as the immunogen
          View immunogen

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Lyophilized

          Concentration

          Conc. Not Determined

          Storage buffer

          whole serum

          Contains

          no preservative

          Storage conditions

          Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Glycerol (1:1) may be added for added stability.

          Shipping conditions

          Ambient (domestic); Wet ice (international)

          Product Specific Information

          Reconstitute in 100 µL of sterile water. Centrifuge to remove any insoluble material.

          The antigen is homologous in mouse, human, chicken and zebrafish. Additionally, the antigen shares ~90% identity with the corresponding phosphorylated sequences in TrkB and TrkC.

          Specificity of this antibody: TrkA, TrkB, TrkC.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          Bioinformatics

          Protein Aliases: BDNF-tropomyosine receptor kinase B; BDNF/NT-3 growth factors receptor; ETS related protein-neurotrophic receptor tyrosine kinase fusion protein; ETV6-NTRK3 fusion; gp140trk; GP145-TrkB; GP145-TrkB/GP95-TrkB; GP145-TrkC; High affinity nerve growth factor receptor; neural receptor protein-tyrosine kinase (trkB); neural receptor protein-tyrosine kinase (trkC); Neurotrophic tyrosine kinase receptor type 1; Neurotrophic tyrosine kinase receptor type 2; Neurotrophic tyrosine kinase receptor type 3; neurotrophic tyrosine kinase, receptor, type 1; neurotrophic tyrosine kinase, receptor, type 2; neurotrophic tyrosine kinase, receptor, type 3; neurotrophic tyrosine receptor kinase type 2; neurotrophin-3; NT-3 growth factor receptor; Oncogene TRK; p140-TrkA; Slow nerve growth factor receptor; trk; Trk-A; Trk-B; Trk-C; TRK1-transforming tyrosine kinase protein; TrkA neurotrophin receptor; trkA proto-oncogene receptor; TrkB tyrosine kinase; TrkC tyrosine kinase; Tropomyosin-related kinase A; Tropomyosin-related kinase B; Tyrosine kinase receptor; Tyrosine kinase receptor A; tyrosine kinase receptor B; tyrosine kinase receptor C

          View more View less

          Gene Aliases: AW125844; C80751; gp145(trkC); GP145-TrkB; GP145-TrkB/GP95-TrkB; GP145-TrkC; MTC; NTRK1; NTRK2; NTRK3; Ntrk3_tv3; p140-TrkA; RATTRKB1; Tkr; Tkrb; TRK; Trk-A; trk-B; TRK1; TRKA; TRKB; TRKB1; TRKC

          View more View less

          UniProt ID: (Human) P04629, (Human) Q16620, (Human) Q16288, (Mouse) Q3UFB7, (Rat) P35739, (Mouse) P15209, (Rat) Q63604, (Mouse) Q6VNS1, (Rat) Q03351

          View more View less

          Entrez Gene ID: (Human) 4914, (Human) 4915, (Human) 4916, (Mouse) 18211, (Rat) 59109, (Mouse) 18212, (Rat) 25054, (Mouse) 18213, (Rat) 29613

          View more View less

          Function(s)
          transmembrane receptor protein tyrosine kinase activity GPI-linked ephrin receptor activity neurotrophin p75 receptor binding protein binding ATP binding nerve growth factor receptor activity kinase binding protein homodimerization activity neurotrophin binding nerve growth factor binding receptor tyrosine kinase binding brain-derived neurotrophic factor binding brain-derived neurotrophic factor-activated receptor activity p53 binding neurotrophin receptor activity nucleotide binding protein kinase activity protein tyrosine kinase activity kinase activity transferase activity transferase activity, transferring phosphorus-containing groups ephrin receptor binding
          Process(es)
          protein phosphorylation transmembrane receptor protein tyrosine kinase signaling pathway multicellular organism development nervous system development axon guidance learning or memory programmed cell death involved in cell development response to activity phosphorylation peptidyl-tyrosine phosphorylation sensory perception of pain cell differentiation B cell differentiation response to nutrient levels response to nicotine nerve growth factor signaling pathway mechanoreceptor differentiation response to drug positive regulation of programmed cell death negative regulation of neuron apoptotic process positive regulation of angiogenesis protein autophosphorylation neurotrophin TRK receptor signaling pathway sympathetic nervous system development detection of temperature stimulus involved in sensory perception of pain detection of mechanical stimulus involved in sensory perception of pain positive regulation of NF-kappaB transcription factor activity response to electrical stimulus positive regulation of synapse assembly positive regulation of synaptic transmission, glutamatergic Sertoli cell development innervation axonogenesis involved in innervation behavioral response to formalin induced pain cellular response to nicotine cellular response to growth factor stimulus negative regulation of neuron death cellular response to nerve growth factor stimulus vasculogenesis neuron migration neuromuscular junction development learning long-term memory feeding behavior positive regulation of cell proliferation positive regulation of gene expression positive regulation of neuron projection development glutamate secretion positive regulation of phosphatidylinositol 3-kinase signaling regulation of metabolic process central nervous system neuron development cerebral cortex development neuron differentiation brain-derived neurotrophic factor receptor signaling pathway positive regulation of peptidyl-serine phosphorylation calcium-mediated signaling using intracellular calcium source neurotrophin signaling pathway regulation of GTPase activity regulation of MAPK cascade positive regulation of MAPK cascade retinal rod cell development regulation of neurotransmitter secretion oligodendrocyte differentiation peripheral nervous system neuron development positive regulation of axonogenesis regulation of dendrite development regulation of protein kinase B signaling retina development in camera-type eye long-term synaptic potentiation cellular response to amino acid stimulus positive regulation of glucocorticoid receptor signaling pathway negative regulation of anoikis activation of MAPK activity negative regulation of protein phosphorylation positive regulation of cell migration activation of protein kinase B activity positive regulation of apoptotic process neuron fate specification positive regulation of axon extension involved in regeneration negative regulation of astrocyte differentiation positive regulation of positive chemotaxis negative regulation of cell death lens fiber cell differentiation activation of GTPase activity positive regulation of actin cytoskeleton reorganization positive regulation of protein phosphorylation inflammatory response aging circadian rhythm response to light stimulus response to auditory stimulus cellular response to tumor necrosis factor cellular response to brain-derived neurotrophic factor stimulus heart development modulation by virus of host transcription response to ethanol ephrin receptor signaling pathway response to axon injury response to corticosterone cellular response to retinoic acid cochlea development negative regulation of cell proliferation response to radiation olfactory nerve development positive regulation of Ras protein signal transduction response to hydrostatic pressure positive regulation of ERK1 and ERK2 cascade activation of MAPKK activity microtubule-based movement positive regulation of GTPase activity phosphatidylinositol-mediated signaling
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-845544bffd-zxsgg:80/100.66.135.11:80.
          git-commit: 2bb44be0f35b85de1c1d8e4f377b85e7a3a71488
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.32.0-2025.07.28-1.0