Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Bioss
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promotions']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.viewpromo']}}
{{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.promocode']}}: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Exhibits a cytosolic function in lipogenesis, adipogenic differentiation, and lipid homeostasis by increasing the activity of ACLY, possibly preventing its dephosphorylation. May act as a transcriptional repressor. Down-regulates CA9 expression.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: ATPase MORC2; ATPase MORC2A; microrchidia 2A; MORC family CW-type zinc finger protein 2; MORC family CW-type zinc finger protein 2A; morc2a; Zinc finger CW-type coiled-coil domain protein 1; zinc finger, CW-type with coiled-coil domain 1
Gene Aliases: 8430403M08Rik; CMT2Z; KIAA0852; MORC2; Morc2a; ZCW3; ZCWCC1
UniProt ID: (Human) Q9Y6X9, (Mouse) Q69ZX6
Entrez Gene ID: (Human) 22880, (Rat) 289736, (Mouse) 74522
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support