Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search Thermo Fisher Scientific

Search All
Search button Close
          • Order Status
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central

          Disclaimer

          Clicking the images or links will redirect you to a website hosted by BenchSci that provides third-party scientific content. Neither the content nor the BenchSci technology and processes for selection have been evaluated by us; we are providing them as-is and without warranty of any kind, including for use or application of the Thermo Fisher Scientific products presented.

          • Primary Antibodies ›
          • gamma Catenin Antibodies

          Invitrogen

          Phospho-gamma Catenin (Tyr550) Polyclonal Antibody

          View all (45) gamma Catenin antibodies

          Promotion

          View available promotion(s)

          Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}

          Datasheet
          Protocols
          Questions & Answers
          Datasheet
          Protocols
          Questions & Answers

          Cite Phospho-gamma Catenin (Tyr550) Polyclonal Antibody

          • Antibody Testing Data (2)
          Phospho-gamma Catenin (Tyr550) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.
          Phospho-gamma Catenin (Tyr550) Antibody in Immunocytochemistry (ICC/IF)
          Group 53 Created with Sketch.

          FIGURE: 1 / 2

          {{ $ctrl.currentElement.advancedVerification.fullName }}
          {{ $ctrl.currentElement.advancedVerification.text }}

          Phospho-gamma Catenin (Tyr550) Antibody (PA5-143668) in ICC/IF

          Immunocytochemistry analysis of Phospho-gamma Catenin (Tyr550) in control (Top) and pervanadate-treated (Bottom) A431 cells. The cells were co-labeled with mouse monoclonal gamma-Catenin antibody or rabbit Phospho-gamma Catenin (Tyr550) polyclonal antibody (Product # PA5-143668), then the antibodies were detected using appropriate secondary antibodies conjugated to Cy2 or Cy3. {{ $ctrl.currentElement.advancedVerification.fullName }} validation info. View more
          Published figure supplied by benchsci-logo
          PMID: {{$ctrl.currentElement.benchSciPubmedId}}
          View Product

          {{$ctrl.videoDesc}}

          {{$ctrl.videoLongDesc}}

          Phospho-gamma Catenin (Tyr550) Antibody in Immunocytochemistry (ICC/IF)
          Phospho-gamma Catenin (Tyr550) Antibody in Western Blot (WB)

          Product Details

          PA5-143668

          Applications
          Tested Dilution
          Publications

          Western Blot (WB)

          1:1,000
          -

          Immunocytochemistry (ICC/IF)

          1:50
          -
          Product Specifications

          Species Reactivity

          Human, Mouse, Rat

          Host/Isotype

          Rabbit / IgG

          Class

          Polyclonal

          Type

          Antibody

          Immunogen

          Phospho-gamma-Catenin (Tyr-550) synthetic peptide (coupled to KLH) corresponding to amino acid residues around tyrosine 550 of human gamma-Catenin. This peptide sequence is highly conserved in rat and mouse gamma-Catenin.

          Conjugate

          Unconjugated Unconjugated Unconjugated

          Form

          Liquid

          Concentration

          0.1 mg/mL

          Purification

          Antigen affinity chromatography

          Storage buffer

          PBS with 1mg/mL BSA, 50% glycerol

          Contains

          0.05% sodium azide

          Storage conditions

          -20°C, Avoid Freeze/Thaw Cycles

          Shipping conditions

          Wet ice

          RRID

          AB_2942896

          Product Specific Information

          This antibody was cross-adsorbed to both phospho-tyrosine coupled to agarose and to dephosphorylated gamma-Catenin (Tyr-550) peptide before affinity purification using phospho-gamma-Catenin (Tyr-550) peptide (without carrier). The antibody detects an 84kDa* protein corresponding to the molecular mass of gamma-Catenin on SDS-PAGE immunoblots of A431 and Hct116 src transformed cells treated with pervanadate (1 mM for 30 min), but not in control cells.

          Target Information

          JUP is a major cytoplasmic protein which is the only known constituent common to sub membranous plaques of both desmosomes and intermediate junctions. This protein forms distinct complexes with cadherins and desmosomal cadherins and is a member of the catenin family since it contains a distinct repeating amino acid motif called the armadillo repeat.

          For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.

          References (0)

          Have you cited this product in a publication?
          Let us know so we can reference it here.
          Cite this product

          Bioinformatics

          Protein Aliases: catenin (cadherin-associated protein), gamma 80kDa; Catenin gamma; Desmoplakin III; Desmoplakin-3; gamma-catenin; gamma-catenin (plakoglobin); Junction plakoglobin; OTTHUMP00000164735

          View more View less

          Gene Aliases: ARVD12; CTNNG; D930025P04Rik; DP3; DPIII; JUP; PDGB; PG; PKGB

          View more View less

          UniProt ID: (Human) P14923, (Rat) Q6P0K8, (Mouse) Q02257

          View more View less

          Entrez Gene ID: (Human) 3728, (Rat) 81679, (Mouse) 16480

          View more View less

          Function(s)
          transcription coactivator activity signal transducer activity structural molecule activity structural constituent of cell wall protein binding protein kinase binding protein phosphatase binding protein homodimerization activity alpha-catenin binding cadherin binding cell adhesion molecule binding cell adhesive protein binding involved in bundle of His cell-Purkinje myocyte communication cadherin binding involved in cell-cell adhesion nuclear hormone receptor binding
          Process(es)
          desmosome assembly cytoskeletal anchoring at plasma membrane signal transduction single organismal cell-cell adhesion cell migration adherens junction organization adherens junction assembly regulation of cell proliferation positive regulation of protein import into nucleus skin development detection of mechanical stimulus positive regulation of sequence-specific DNA binding transcription factor activity protein heterooligomerization endothelial cell-cell adhesion cellular response to indole-3-methanol bundle of His cell-Purkinje myocyte adhesion involved in cell communication regulation of heart rate by cardiac conduction establishment of protein localization to plasma membrane positive regulation of canonical Wnt signaling pathway regulation of ventricular cardiac muscle cell action potential cell adhesion regulation of cell fate specification positive regulation of transcription from RNA polymerase II promoter ventricular cardiac muscle cell action potential bundle of His cell to Purkinje myocyte communication regulation of nucleic acid-templated transcription
          It has to be done as per old AB suggested Products section.
          guarantee_icon

          Performance Guarantee

          If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*

          Learn more
          help_icon

          We're here to help

          Get expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.

          Contact tech support
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us
          • Careers
          • Investors
          • News
          • Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-5d94444789-k2m5x:80/100.66.128.182:80.
          git-commit: ec3573d8beaf0fd412f2d0ea2dcb8dd14c74098c
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.31.0-2025.06.23-1.0