Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Invitrogen
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Antibody detects endogenous levels of p73 only when phosphorylated at Ser388.
P73 (TP73) participates in the apoptotic response to DNA damage. Isoforms containing the transactivation domain are pro-apoptotic, isoforms lacking the domain are anti-apoptotic and block the function of p53 and transactivating p73 isoforms. May be a tumor suppressor protein.
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: OTTHUMP00000003479; OTTHUMP00000003480; p53-like transcription factor; p53-related protein; RP23-254N4.10; Tumor protein p73
Gene Aliases: deltaNp73; P73; TAp73; TP73; Trp73
UniProt ID: (Human) O15350, (Mouse) Q9JJP2
Entrez Gene ID: (Human) 7161, (Mouse) 22062
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support