Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Proteintech
Promotion
View available promotion(s)
Promo code: {{promo.promoCode}} {{promo.promoDescription}}. {{$productOrderCtrl.translations['antibody.pdp.commerceCard.promotion.learnmore']}}
Immunogen sequence: EKQQDSQPE EVMDVLEMVE NVKHVIADQE VMETNRVESV EPSENEASKE LEPEMEFEIE PDKECKSLSP GKENVSALDM EKESEEKEEK ESEPQPEPVA QPQPQSQPQL QLQSQSQPVL QSQPPSQPED LSLAVLQPTP QVTQEQGHLL PERKDFPVES VKLTEVPVEP VLTVHPESKS KTKTRSRSRG RARNKTSKSR SRSSSSSSSS SSSTSSSSGS SSSSGSSSSR SSSSSSSSTS GSSSRDSSSS TSSSSESRSR SRGRGHNRDR KHRRSVDRKR RDTSGLERSH KSSKGGSSRD TKGSKDKNSR SDRKRSISES SRSGKRSSRS ERDRKSDRKD KRR (376-717 aa encoded by BC062602)
Transcriptional activator binding to the E-box 1 core sequence of the E-cadherin promoter gene; the core-binding sequence is 5'CAGGTG-3'. Capable of reversing CTBP1-mediated transcription repression. Auxiliary component of the splicing-dependent multiprotein exon junction complex (EJC) deposited at splice junction on mRNAs. The EJC is a dynamic structure consisting of core proteins and several peripheral nuclear and cytoplasmic associated factors that join the complex only transiently either during EJC assembly or during subsequent mRNA metabolism. Participates in the regulation of alternative pre-mRNA splicing. Associates to spliced mRNA within 60 nt upstream of the 5'-splice sites. Component of the PSAP complex which binds RNA in a sequence-independent manner and is proposed to be recruited to the EJC prior to or during the splicing process and to regulate specific excision of introns in specific transcription subsets. Involved in the establishment and maintenance of epithelia cell-cell adhesion. Potential tumor suppressor for renal cell carcinoma. [UniProt]
For Research Use Only. Not for use in diagnostic procedures. Not for resale without express authorization.
Protein Aliases: 140 kDa nuclear and cell adhesion-related phosphoprotein; Desmosome-associated protein; Domain-rich serine protein; DRS protein; Melanoma metastasis clone A protein; neutrophil protein; Nuclear protein SDK3; Pinin; SR-like protein
Gene Aliases: DRS; DRSP; MEMA; PNN; SDK3
UniProt ID: (Human) Q9H307
Entrez Gene ID: (Human) 5411
If an Invitrogen™ antibody doesn't perform as described on our website or datasheet,we'll replace the product at no cost to you, or provide you with a credit for a future purchase.*
Learn moreGet expert recommendations for common problems or connect directly with an on staff expert for technical assistance related to applications, equipment and general product use.
Contact tech support